Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001147614.1 | NP_001141086.1 | 1024 | 81 | ||
GenBank mRNA | BT040366.1 | - | 1024 | 81 | ||
BT085766.1 | - | 990 | 81 | |||
BT088241.1 | - | 904 | 81 |
Gene Symbol: LOC100381348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001174213.1 | NP_001167684.1 | 916 | 81 | ||
GenBank mRNA | EU961218.1 | - | 887 | 81 | ||
EU964551.1 | - | 916 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Zea mays |