Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001147950.1 | NP_001141422.1 | 767 | 59 | ||
GenBank mRNA | BT041345.1 | - | 767 | 59 | ||
BT063632.1 | - | 767 | 59 | |||
BT069295.1 | - | 734 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Zea mays |