Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112174.1 | NP_001105644.1 | 651 | 65 | ||
GenBank mRNA | AF366295.1 | - | 651 | 65 | ||
AY109666.1 | - | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Zea mays |