Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112175.1 | NP_001105645.1 | 731 | 59 | ||
GenBank mRNA | AF370003.1 | - | 731 | 59 | ||
BT064355.1 | - | 675 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Zea mays |