Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156240.1 | NP_001149712.1 | 324 | 68 | ||
GenBank mRNA | BT041981.1 | - | 171 | 68 | ||
EU961092.1 | - | 201 | 68 | |||
EU964366.1 | - | 324 | 68 | |||
EU976212.1 | - | 330 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Zea mays |