Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111536.1 | NP_001105006.1 | 535 | 86 | ||
GenBank mRNA | AF283564.2 | - | 535 | 86 | ||
EU967840.1 | - | 604 | 86 |