Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111627.1 | NP_001105097.1 | 748 | 106 | ||
GenBank mRNA | AF105149.1 | - | 352 | 106 | ||
AF529266.1 | - | 748 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Zea mays |