Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111892.1 | NP_001105362.1 | 424 | 81 | ||
GenBank mRNA | AY111119.1 | - | 424 | 81 | ||
EU961361.1 | - | 545 | 81 | |||
U10076.1 | - | 424 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Zea mays |