Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sum1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112254.1 | NP_001105724.1 | 562 | 71 | ||
GenBank mRNA | D83391.1 | - | 562 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Zea mays |