Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112255.1 | NP_001105725.1 | 422 | 62 | ||
GenBank mRNA | X65931.1 | - | 422 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Zea mays |