Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pti1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112283.1 | NP_001105753.1 | 555 | 77 | ||
GenBank mRNA | AY554281.1 | - | 555 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Zea mays |