Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP1-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111552.1 | NP_001105022.1 | 1152 | 98 | ||
GenBank mRNA | AF326487.1 | - | 1152 | 98 | ||
BT040812.1 | - | 96 |
Set Membership: |
Amplicon length between 86 and 100 Transporter 3' Most Recommended Assay Zea mays |