Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112301.1 | NP_001105771.1 | 250 | 79 | ||
GenBank mRNA | AY850139.1 | - | 250 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Zea mays |