Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nbp35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001155963.1 | NP_001149435.1 | 1654 | 100 | ||
GenBank mRNA | BT043183.1 | - | 1588 | 100 | ||
EU963318.1 | - | 1654 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Zea mays |