Search
| Catalog Number | Cooling Capacity | Temperature Range | Model |
|---|---|---|---|
| TF900 | Up to 900 watts (60 Hz) 750 watts (50 Hz) | 5°C to 40°C | ThermoFlex 900 |
| TF900-LN | Up to 900 watts (60 Hz) 750 watts (50 Hz) | 5°C to 40°C | ThermoFlex 900 Low Noise |
| TF1400 | Up to 1400 watts (60 Hz) 1170 watts (50 Hz) | 5°C to 90°C | ThermoFlex 1400 |
| TF1400-LN | Up to 1400 watts (60 Hz) 1170 watts (50 Hz) | 5°C to 40°C | ThermoFlex 1400 Low Noise |
| TF2500 | Up to 2500 watts (60 Hz) 2200 watts (50 Hz) | 5°C to 90°C | ThermoFlex 2500 |
| TF3500 | Up to 3500 watts (60 Hz) 3050 watts (50 Hz) | 5°C to 90°C | ThermoFlex 3500 |
| TF5000 | Up to 5000 watts (60 Hz) 4400 watts (50 Hz) | 5°C to 90°C | ThermoFlex 5000 |
| TF7500 | Up to 7500 watts (60 Hz) 6425 watts (50 Hz) | 5°C to 90°C | ThermoFlex 7500 |
| TF10000 | Up to 10,000 watts (60 Hz) 8500 watts (50 Hz) | 5°C to 90°C | ThermoFlex 10000 |
| TF15000 | Up to 15,000 watts (60 Hz) 12,525 watts (50 Hz) | 5°C to 40°C | ThermoFlex 15000 |
| TF20000 | Up to 20,000 watts (60 Hz) 16,700 watts (50 Hz) | 5°C to 40°C | ThermoFlex 20000 |
| TF24000 | Up to 24,000 watts (60 Hz) 21,000 watts (50 Hz) | -5°C to +50°C | ThermoFlex 24000 |