Search
Applied Biosystems CopyCaller Software simplifies the analysis of copy number data from real-time PCR experiments using TaqMan Copy Number Assays. Raw data are collected and relative quantitation analysis is performed with CopyCaller Software to determine the copy number of each genomic DNA target.
CopyCaller Software does the following:
The updated version of CopyCaller Software, version 2.1, has been released for download.
Updated content includes:
Additional instrument compatibility
CopyCaller Software v2.1 can import copy number assay results files generated using QuantStudio 6, 7, and 12K Flex Systems.
Additional analysis capability
CopyCaller Software v2.1 can analyze data generated by the Crt “relative threshold” method (available on QuantStudio 6, 7, and 12K Flex Systems and ViiA 7™ Real-Time PCR System) in addition to the Ct “baseline threshold” method.
Note: Minor bug fixes were also implemented. A complete list of features and fixes can be found in the Read Me file.
PC Download CopyCaller™ Software v2.1 264 MB Read Me | User Guide
For Research Use Only. Not for use in diagnostic procedures.