Search
We are dedicated to your success. Get back on track. View our expert recommendations for commonly encountered problem scenarios.
View the relevant questions below:
Beginning your experiment? Visit our
The fading is most likely due to detergent in the western blocking/washing solutions that can remove some of the proteins from the membrane. The dye itself will not wash off of the proteins because it is covalently bound. We have found that smaller pore size membranes retain the proteins better during blocking and wash procedures, and hence recommend use of 0.2 µm instead of 0.45 µm membranes for best resolution and protein retention. After transfer, it is a good idea to circle the pre-stained bands with a pencil on the membrane, so band positions can be identified after blocking and processing.
Here are possible causes and solutions:
| Cause | Solution |
| The membrane may not be properly treated prior to transfer | Make sure that the membrane is pre-wetted with a polar organic solvent such as methanol or ethanol. |
| There may be poor gel to membrane contact | Ensure that the filter paper and blotting pads are well saturated with transfer buffer, taking care to remove any bubbles during the assembly of the membrane sandwich. The gel/membrane sandwich must fit securely in the two halves of the blot module. Try adding another pad or replace any pads that have lost their resiliency with fresh ones. |
| Over-compression of the gel | A good indication of over-compression is if the gel has been excessively flattened. In the event that the sandwich is over-compressed, remove enough pads so that the blotter can be closed without exerting excess pressure on the gel and the membrane. |
Here are possible causes and solutions:
| Cause | Solution |
| The buffer was accidentally made too dilute, therefore increasing resistance and thus lowering conductivity and current | Check the transfer buffer and its reagent components and then re-dilute it or remake it. |
| The circuit is broken or impeded, as in the case of a corroded or broken electrode or malfunctioning power supply | Check the equipment. |
| There is a leak in the blot module (this is indicated by a drastic decrease in current and in buffer volume within the module) | Ensure that the inner buffer chamber is filled sufficiently so that the wells are covered with buffer. |
| Tape at the bottom of the gel cassette was not removed | Double check that the tape on the bottom of the gel has been removed. |
The most common cause of abnormally high current is the buffer. If the buffer is too concentrated, this leads to increased conductivity and higher current. High current may also occur if Tris-HCl was accidentally substituted for the Tris base required in the transfer buffer. Tris-HCl results in a low buffer pH and leads to increased conductivity and current, and, subsequently, overheating. Check the transfer buffer and its reagent components, re-dilute, or remake the buffer.
We recommend discarding the buffer and remaking it after rechecking the reagents and the water purity. We do not recommend adjusting the pH with acid or base, as this will increase the conductivity of the buffer and result in higher current during the transfer.
It is possible that the gel/membrane sandwich was assembled in the reverse direction such that the proteins have migrated out into the buffer. Assemble the blot sandwich in the correct order using instructions provided in the manual.
Here are possible causes and solutions:
| Cause | Solution |
| Too long a transfer time | Shorten the transfer time by 15 minute increments. |
| Inappropriate amount of SDS | Do not include any SDS in the transfer buffer. |
| Inappropriate methanol content | Add additional methanol to the transfer buffer to increase the binding capacity of the membrane. |
| Inappropriate gel type | Check the percentage of the gel used and switch to a higher percentage gel. |
| Sample overloaded | Decrease the sample load. |
| Finally, if using nitrocellulose membrane, switch to PVDF which has a higher binding capacity. |
Here are possible causes and solutions:
Cause | Solution |
Too short a transfer time | Increase the blotting time by 15 minute increments. |
Inappropriate gel type | Check the percentage of the gel used and switch to a higher percentage gel. |
Inappropriate amount of SDS | Add 0.01–0.02% SDS to the transfer buffer to facilitate migration of the protein out of the gel. |
Inappropriate methanol content | Decrease the amount of methanol in the transfer buffer. |
Note: Higher molecular weight proteins usually do not transfer completely as compared to mid to low molecular weight proteins.
Here are possible causes and solutions:
The swirling and diffuse banding patterns are typical of molecules moving laterally before binding to the membrane during transfer. Here are possible causes and solutions:
Cause | Solution |
Poor contact between the gel and the membrane | The gel should be attached to the membrane through capillary action. To ensure that this happens, make sure that you roll over the surface of each layer of the gel/membrane sandwich with a glass pipette to ensure good contact between the gel and the membrane. It is helpful to use a disposable pipette to place some extra transfer buffer on the surface of each layer as the sandwich is being made. Also, the pads need to be fully saturated (push down with gloved hand when they are placed in transfer buffer to make sure there are no air bubbles.) |
Under-compression of the gel | The gel/membrane assembly should be held securely between the two halves of the blot module. Try adding another pad or replace any pads that have lost their resiliency with fresh ones. |
Over-compression of the gel | A good indication of over-compression is if the gel has been excessively flattened. In the event that the sandwich is over-compressed, remove enough pads so that the blotter can be closed without exerting excess pressure on the gel and membrane. |
Note: The height of the uncompressed pads should be 0.5–1.0 cm above the level of the sealing gasket.
Here are possible causes and solutions:
For proteins larger than 100 kDa, we recommend pre-equilibrating the gel in 2X NuPAGE™ Transfer buffer (without methanol) containing 0.02–0.04% SDS for 10 minutes before assembling the sandwich and then transferring using 1X NuPAGE™ transfer buffer containing methanol and 0.01% SDS.
The most common cause of abnormally high current is the transfer buffer. If the transfer buffer is too concentrated, this leads to increased conductivity and current. High current may also occur if Tris-HCl is accidentally substituted for the Tris base required in the transfer buffer. This will again result in low buffer pH and lead to increased conductivity and current and subsequently, overheating. We recommend checking the transfer buffer and its reagent components and re-diluting or remaking the buffer.
Here are possible causes and solutions:
| Cause | Solution |
| Voltage is too low | 1-mm thick polyacrylamide gels (Mini and Midi Gels) should be transferred at 20 V, E-PAGE™ Gels at 25 V (approximately 15 V/cm field strength). |
| Power supply is inappropriate for semidry transfer | Some power supplies will shut off or blow a fuse when run at the conditions required for semi-dry transfer. Semi-dry transfer requires low voltage (20 V) and high current. Check with the manufacturer of the power supply to determine whether it is appropriate for semi-dry transfer. |
| Transfer performed for too short a time | Increase the amount of time for transfer. Typical transfer times range from 30 to 60 minutes. |
| Transfer sandwich was assembled in the wrong order | The Novex™ Semi-dry Blotter is configured with the cathode on the top, and anode on the bottom. This results in a downward transfer of proteins from the gel onto the membrane. Follow the instructions carefully when assembling the transfer sandwich. |
| The pH of the transfer buffer is too close to the isoelectric point of the protein | The transfer buffers should be at the optimal pH if prepared as described in this manual. Do not adjust the pH with acid or base as this will increase the conductivity of the buffer and result in higher current during transfer. |
| Too much methanol in the transfer buffer | Reducing the amount of methanol can help elute proteins from the gel, but can reduce binding to nitrocellulose membranes. |
| High-percentage gels restrict transfer | Higher percentage acrylamide or crosslinkers can restrict elution of proteins. Use the lowest percentage acrylamide possible to separate your proteins. |
| Puddles of buffer were present on the plates, allowing the current to bypass the stack | Always clean up the lower plate before closing the lid of the transfer apparatus. Do not squeeze the stack excessively, as this removes transfer buffer from the blotting paper and also creates puddles that the current can pass through. |
Here are possible causes and solutions:
| Cause | Solution |
| Air spaces are interfering with contact between the gel and the membrane | Roll the membrane with a blotting roller (or a clean test tube or pipet) before placing the gel on the membrane, and then remove any air bubbles between the gel and membrane with a blotting roller or a wet gloved finger. Transfer will not occur where the gel is not in contact with the membrane. |
| Electrophoretic conditions were incorrect or not ideal | Running conditions, sample preparation, percentage acrylamide, and many other variables can affect the migration and resolution of proteins. Please review your electrophoresis conditions. |
| Under- or over-compression of gel | Follow the compression guidelines in the manual. Too little compression can allow proteins to migrate between the gel and membrane, causing protein band smearing. Too much compression can distort the gel. |
| Cause | Solution |
| Over-transfer through the membrane | Use 0.2 μm pore size nitrocellulose instead of 0.45 μm, or use PVDF with a higher binding capacity. |
| Not enough methanol in the transfer buffer | Increase the concentration of methanol. |
| Low molecular weight proteins are not binding well or are being washed away | Use glutaraldehyde to crosslink the proteins to the membrane and use Tween™ 20 in the wash steps. |
| SDS is included in the transfer buffer | Do not use SDS in the transfer buffer. |
Here are possible causes and solutions:
Here are possible causes and solutions:
This indicates an incomplete electric circuit and here are possible causes and solutions:
| Cause | Solution |
| iBlot™ Disposable Sponge covers the metal contact or the metal contact on the sponge is on the left | Reinsert the iBlot™ Disposable Sponge such that the metal contact on the sponge is on the top right of the lid and is in contact with the electrode on the transfer stack. |
| Incorrect position of the transfer stacks or improper assembly of the transfer stacks | Make sure the transfer stack is placed in the proper position in the blotting surface to allow proper contacts with the electrodes. Ensure the transfer stacks are assembled correctly; use the iBlot™ Anode Stack, Bottom first followed by the gel and iBlot™ Cathode Stack, Top. |
| Incorrect position of the pull tab | Ensure the pull tab from the iBlot™ Cathode Stack, Top is towards the right of the assembly in the blotting surface. |
| iBlot™ Anode Stack, Bottom placed on the device without the tray including the electrical contact | Do not remove the iBlot™ Anode Stack, Bottom from the tray during the assembly. The blotting is performed with the bottom stack in the plastic tray. |
| iBlot™ Cathode Stack, Top placed on the device with the tray | Always remove the iBlot™ Cathode Stack, Top from the red plastic tray before placing the top stack on the assembly. Do not use the iBlot™ Cathode Stack, Top with the tray. |
| The metal safety contacts in the lid hinge may be dirty and do not make contact | Clean the metal safety contacts in the lid hinge with a cotton swab and water. |
This indicates an open electrical circuit during the run. It occurs if the lid opened during the run.
Close the lid and continue the run by briefly pressing the Start/Stop Button or restart the run by pressing and holding the Start/Stop Button.
| Cause | Solution |
| The iBlot™ Cathode Stack, Top is touching the copper electrode on the iBlot™ Anode Stack, Bottom | Open the lid and align the iBlot™ Cathode Stack, Top to the right. Continue the run by briefly pressing Start/Stop Button or restart the run by pressing and holding the Start/Stop Button. |
| The layers are not aligned | Align the layers properly as described in the protocols. Ensure that the electrodes are in contact. |
| Current is above 5.5 amp | Select a program with a lower voltage. Open the iBlot™ Lid and ensure the stacks are aligned properly. Close the lid and restart the run by subtracting the time already elapsed. Replace the iBlot™ Gel Transfer Stacks with fresh transfer stacks. Ensure an iBlot™ Filter Paper was used during blotting of mini- or midi-gels. |
This is most likely due to incorrect placement of the top Stack. Be sure the iBlot™ Cathode Stack, Top is placed correctly with the copper electrode side facing up. Avoid placing the top stack in the inverted position.
Longer transfer times result in the deposition of copper ions, causing the blue discoloration. Be sure to perform the transfer for the recommended time for each gel type.
The green discoloration is due to copper ions from the iBlot™ Transfer Stacks being carried by liquids and deposited on the membrane. These deposits do not interfere with downstream processes. The stained regions can be cut away, but membrane washing typically results in their removal. To minimize this effect, shake excess water off the filter paper and buffer from the gel before placing each on the stack.
This happens if the membrane is trimmed to fit the gel size resulting in direct contact between the iBlot™ Cathode, Top and Anode Bottom stacks. Always maintain the membrane size identical to the transfer stack. Transfer quality is not affected by smaller gel size compared to the membrane.
This could be due to no current passing through or because an incorrect voltage Method was used. Make sure that the electrical circuit is complete and current is flowing through the device. Please check to make sure that the correct voltage Method is used (see page 13 in the manual).
Here are possible causes and solutions:
This could happen if an incorrect voltage Method was used or if inappropriate transfer conditions were used. Make sure that the voltage Method and run time used is correct, based on the gel type, as described on page 13 in the manual.
For mini or midi gels:
For E-PAGE™ gels:
Note: It is normal for some proteins to remain in the gel because some high–molecular weight proteins do not transfer completely using the iBlot™ Gel Transfer Device, compared to semi-wet transfer apparatus.
This could happen if the transfer time used is too long. We recommend reducing the transfer time by 30 second increments.
Note: Pre-stained markers are charged, so tend to blow-through more than regular proteins.
This indicates that a non-uniform electric field was created around the wells. Ensure that the well protrusions on the E-PAGE™ gel are properly flattened using the De-bubbling Roller. To ensure the best blotting results, we recommend using the De-bubbling Roller with E-PAGE™ gels. If you use the Blotting Roller with E-PAGE™ gels, be sure to follow the recommendations on page 22 of the manual to obtain good results.
This is likely due to the PVDF membrane being dry or partially dry. Regions where PVDF membranes are dry appear whiter than places where the membrane is wet. Remove the membrane and reactivate in 100% methanol, and rinse in water before reapplying to the transfer stack.
This is likely due to use of TBST buffers for washing. We recommend using PBST or WesternBreeze™ wash solutions.
This is most likely due to the protein load being too high, as a result of which detection is not within the linear range. Since the immunodetection sensitivity is higher for dry blotting with the iBlot™ Gel Transfer Device than for semi-dry or wet blotting, we recommend that you decrease the protein load, use more diluted antibody, or perform detection for shorter time. You may need to perform some optimization based on your initial results.
It is possible that the PVDF membrane got displaced during handling or shipment. The activated PVDF membrane is transparent, making it difficult to see. If it is not present on top of the stack, we recommend examining the aluminum seal of the Anode Stack (clear tray) to make sure that the PVDF membrane has not adhered to the seal. If the membrane has adhered to the seal, we suggest reactivating the membrane with pure methanol, then rinsing it well in distilled water, and carefully placing it on the stack. The performance will not be affected by the reactivation process.
The iBlot™ device must be connected directly to the computer for the firmware upgrade to work. Do not use a USB hub.
In order to make a loop-back test you first need to make sure that the drivers and COM port have been successfully installed.
If the above steps don’t help, another simple option that may solve the issue would be to try a different USB cable.
This could happen due to the USB cable disconnecting during the updating process. Please try again with another computer and a different USB cable.
This is most likely due to uneven current distribution during transfer, possible caused by bad electrodes. We recommend getting new electrodes (Cat. No. IB1002).
This indicates an incomplete electric circuit. Here are possible causes and solutions:
| Cause | Solution |
| Incorrect placement of iBlot™ 2 Absorbent Pad contact | Make sure the electrical contact of the iBlot™ 2 Absorbent Pad is aligned with the corresponding electrical contacts on the blotting surface of the iBlot™2 Gel Transfer Device. |
| Top Stack placed on the device upside down | Make sure the Top Stack is assembled with the copper electrode facing up. |
| The metal safety contacts in the lid hinge may be dirty and do not make contact | Clean the metal safety contacts in the lid hinge with a cotton swab and water. |
| Plastic separator was not removed when assembling stack | Make sure that the plastic separator is removed from the stack (see page 21 in the manual). |
This could be caused by incorrect placement of the Top Stack. Please check to make sure that the Top Stack is placed correctly with the copper electrode facing up. Avoid placing the top stack in the inverted position.
Longer transfer times result in the deposition of copper ions. Be sure to perform the transfer for the recommended time for each gel type.
The green discoloration is due to copper ions carried with liquids that get deposited onto the membrane. These deposits do not interfere with downstream processes. The stained regions can be cut away, but membrane washing typically results in their removal.
This happens if the membrane is trimmed to fit the gel size resulting in direct contact between the Top and Bottom stacks. This can be avoided by maintaining the membrane size identical to the transfer stack. Transfer quality is not affected by smaller gel size compared to the membrane.
This could be due to no current passing through or because an incorrect voltage Method was used. Make sure that the electrical circuit is complete and current is flowing through the device. Please check to make sure that the correct voltage Method is used (see page 17 in the manual).
Here are possible causes and solutions:
This could happen if an incorrect voltage Method was used or if inappropriate transfer conditions were used. Make sure that the voltage Method and run time used is correct, based on the gel type, as described on page 17 in the manual.
For mini or midi gels:
For E-PAGE™ gels:
Note: It is normal for some proteins to remain in the gel because some high–molecular weight proteins do not transfer completely using the iBlot™ 2 Gel Transfer Device, compared to semi-wet transfer apparatus.
This could happen if the transfer time used is too long. We recommend reducing the transfer time by 30 second increments.
Note: Pre-stained markers are charged, so tend to blow-through more than regular proteins.
This indicates that a non-uniform electric field was created around the wells. Ensure that the gel is properly flattened by using the Blotting Roller. Follow the recommendations on page 19 in the manual to obtain good results.
This is likely due to the PVDF membrane being dry or partially dry. Regions where PVDF membranes are dry appear whiter than places where the membrane is wet. Remove the membrane and reactivate in 100% methanol, and rinse in water before reapplying to the transfer stack.
This is likely due to use of TBST buffers for washing. We recommend using PBST or WesternBreeze™ wash solutions.
This is most likely due to the protein load being too high, as a result of which detection is not within the linear range. Since the immunodetection sensitivity is higher for dry blotting with the iBlot™ 2 Gel Transfer Device than for semi-dry or wet blotting, we recommend that you decrease the protein load, use more diluted antibody, or perform detection for shorter time. You may need to perform some optimization based on your initial results.
This is likely due to salt deposited on moving parts inside the cassette. Rinse the cassette top and bottom under warm water while removing any sticky salt residue with a gloved hand. Briefly rinse with deionized water and place in a rack to dry.
For more thorough cleaning, immerse the unassembled cassette in warm water, use a gloved hand or clean sponge to remove any sticky salt residue. Rinse with deionized water and place perpendicular in a rack to dry.
Note: Failure to keep cassette top (cathode) and bottom (anode) clean will result in the sticking of moving parts and lead to poor transfer efficiency.
Here are possible causes and solutions:
| Cause | Solution |
| Membrane or filter paper was insufficiently equilibrated in Thermo Scientific™ Pierce™ 1-Step Transfer Buffer | Equilibrate membrane and filter paper in Thermo Scientific™ Pierce™ 1-Step Transfer Buffer before transfer for a minimum of 5 minutes. Use a sufficient amount of buffer for the equilibration step. |
| Insufficient transfer time | Increase transfer time from 7–10 minutes to 10–12 minutes. |
| PVDF membrane was not pre-wetted with methanol or ethanol | Wet PVDF membrane with methanol or ethanol and equilibrate for 10–15 minutes in Thermo Scientific™ Pierce™ 1-Step Transfer Buffer before transfer. |
| Air bubbles trapped between gel and membrane | When assembling transfer stack, use a roller or pipette to remove any air bubbles between the gel and the membrane. |
| Filter paper used in the fast transfer exceeded 1.8 mm | Use filter paper <1.8 mm thick. |
This is most likely due to inefficient binding of some low-molecular weight proteins (<25 kDa) to PVDF membrane. We recommend combining ethanol and Thermo Scientific™ Pierce™ 1-Step Transfer Buffer in a 15:85 ratio before equilibrating filter paper and membrane (e.g., 15 mL of ethanol with 85 mL of Thermo Scientific™ Pierce™ 1-Step Transfer Buffer).
PVDF membranes require more stringent blocking steps. This can be achieved by increasing the concentration of the blocking agent 2–5 fold, increasing the blocking time, and performing the procedure at 37 degrees C. Blocking agents bind to unoccupied sites to prevent background staining and also to membrane-bound proteins, reducing non-specific interactions with the primary antibody. Examples of blocking agents are nonfat dry milk, BSA, and casein.
The most probable cause of the swirled artifact is the way the membrane is moved around in the staining solution. Using an orbital shaker can cause this effect. To eliminate these background patterns, we recommend switching to a rocker type shaker or combination of rocker and reciprocal motions to ensure even sloshing of the staining solution over the membrane.
For Research Use Only. Not for use in diagnostic procedures.