Search
Biotherapy manufacturers, especially those producing monoclonal antibodies (mAbs), rely heavily on chromatography for its precision and accuracy. Chromatography is an ideal technology to leverage in maintaining the purity, potency, and safety of these vital therapeutic agents. Techniques like High-Performance Liquid Chromatography (HPLC), Ion-Exchange Chromatography (IEX), and Size-Exclusion Chromatography (SEC) are crucial for detecting impurities, assessing molecular characteristics, and quantifying active ingredients.
Chromatography also helps identify protein aggregates and provides detailed insights into the molecular structure and charge variants of mAbs. This thorough analysis maintains consistency and reproducibility across batches, enhancing product quality and supporting regulatory compliance.
Moreover, chromatography streamlines the manufacturing process. It allows manufacturers to monitor production stages, identify issues early, and make necessary adjustments, thereby improving yield and product quality. Integrating chromatography into analytical applications is essential for manufacturers to deliver high-quality therapeutic agents to the market.
Analytical testing is crucial in the bioprocessing workflow to support the quality and safety of biopharmaceuticals. It helps detect impurities, assess molecular characteristics, and quantify active ingredients, maintaining consistency and reproducibility across batches. This rigorous testing supports regulatory compliance and contributes to overall product quality.
Thermo Fisher Scientific offers a robust portfolio of testing solutions to help simplify analytical processes while helping you meet or exceed regulatory guidelines.
Throughout the manufacturing process, analytical chromatography is employed to assess the molecular characteristics, quantify active ingredients, and support consistency and reproducibility across product batches.
When choosing the right HPLC column for your analytical application, consider several key factors. The molecular size and characteristics must be considered when selecting pore size and surface chemistry to help ensure optimal separation. The material and stability of the column are also crucial, as they need to withstand the mobile phase conditions and remain reliable through repeated use. Additionally, the column's resolution and efficiency are important for achieving accurate and reproducible results. Finally, regulatory compliance and validation requirements should guide the selection process to help ensure that the column meets the necessary standards for use in mAb production and quality control.
Thermo Scientific POROS resins are engineered to deliver fast, high-resolution analytical separations, supporting accurate and reproducible characterization of biomolecules. Built on a robust polymeric matrix, POROS technology offers exceptional mechanical stability with high flow rates, with consistent performance even under demanding HPLC conditions.
The unique bead structure features a large surface area that enables rapid mass transfer, facilitating reliable charge variant analysis, aggregate detection, and molecular profiling—all with greater speed and efficiency. With proven versatility across analytical applications from development through manufacturing, POROS chemistries empower analysts to generate actionable data quickly and with confidence, supporting quality and consistency at every stage.
Attributes of POROS chromatography resins:
Thermo Scientific CaptureSelect affinity ligands offer exceptional specificity and selectivity to analytical workflows, enabling targeted detection and quantification of diverse biologics—from full-length antibodies to antibody fragments and next-generation therapeutics. Built on innovative single-domain antibody technology, CaptureSelect chemistries facilitate precise binding, reproducibility, and efficiency, simplifying complex assays while improving accuracy and reliability.
Well suited for challenging biological samples, CaptureSelect ligands help enhance analytical column performance by facilitating clean, consistent separations and dependable results. When paired with POROS bead technology, they enable highly specific affinity separations and molecular characterization that support the quality, consistency, and scalability required from development through manufacturing.
Benefits of CaptureSelect ligands:
No matter your questions, we are here to help. Our experienced team of scientists and technical specialists is ready to assist you in choosing the right solution and products. Complete this form to start the conversation.
Explore the Analytics Knowledge Hub and discover a robust collection of webinars, articles, eBooks, infographics, and other digital resources to help you optimize analytical testing in your bioprocess workflow.
Discover areas where you can cut costs and gain efficiency using our integrated calculator.
For research and development use only in support of FDA-regulated end uses. Not for diagnostic use or direct administration to humans or animals.