Search
Jurkat (human T-cell leukemia) cells were treated with 10 µM EdU for 2 hours, stained with CD3 mouse anti-human mAb PE-Cy®5.5 (Cat. No. MHCD0318) and detected according to the recommended staining protocol. The figures show a clear separation of proliferating cells that have incorporated EdU and nonproliferating cells that have not. Panel A shows data from cells labeled with Alexa Fluor® 647 picolyl azide analyzed on an Attune® Acoustic Focusing Cytometer using 635 nm excitation and a 660/20 nm bandpass emission filter; Panel B shows the same cells using 488 nm excitation and a 574/26 nm bandpass emission filter. The black outlined histogram is the cells stained with CD3 mouse anti-human mAb PE-Cy®5.5 and Click-iT® Plus EdU Alexa Fluor® 647 picolyl azide. The gray outlined histogram is the CD3 mouse anti-human mAb PE-Cy®5.5 positive control cells treated the same but without copper in the reaction.