Search
ExpressionSuite Software is a free, easy-to-use data-analysis tool that utilizes the comparative Cτ (ΔΔCτ) method to rapidly and accurately quantify relative gene expression across a large number of genes and samples. Its flexibility allows the user to analyze gene expression data on any current Applied Biosystems® real-time PCR instrument. The software showcases an intuitive user interface and enhanced multiplate analysis features to meet the requirements of emerging markets and future research.
Key product features:
You can import real-time PCR data files and create a study for multiplate data analysis from all supported Applied Biosystems™ instruments including the QuantStudio™ 3 Real-Time PCR, QuantStudio™ 5 Real-Time PCR, QuantStudio™ 12K Flex Real-Time PCR, ViiA™ 7 Real-Time PCR, 7500 Fast Real-Time PCR, 7500 Real-Time PCR, StepOnePlus™ Real-Time PCR, StepOne™ Real-Time PCR, and 7900HT Fast Real-Time PCR
Software Installs | |||||
![]() | PC | Download | ExpressionSuite release v1.3 release note | ||
![]() | PC | Download | ExpressionSuite release v1.3 software | ||
![]() | PC | Download | ExpressionSuite for QuantStudio™ 6 & 7 Flex Real Time PCR System |
| |
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use