Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Amino Acids | |||
| Glycine | 75.0 | 7.5 | 0.1 |
| L-Alanine | 89.0 | 9.0 | 0.101123594 |
| L-Arginine hydrochloride | 211.0 | 211.0 | 1.0 |
| L-Asparagine-H2O | 150.0 | 15.0 | 0.1 |
| L-Aspartic acid | 133.0 | 13.0 | 0.09774436 |
| L-Cysteine | 121.0 | 25.0 | 0.20661157 |
| L-Glutamic Acid | 147.0 | 14.7 | 0.1 |
| L-Glutamine | 146.0 | 146.0 | 1.0 |
| L-Histidine hydrochloride-H2O | 210.0 | 23.0 | 0.10952381 |
| L-Isoleucine | 131.0 | 2.6 | 0.019847328 |
| L-Leucine | 131.0 | 13.0 | 0.099236645 |
| L-Lysine hydrochloride | 183.0 | 29.0 | 0.15846995 |
| L-Methionine | 149.0 | 4.5 | 0.030201342 |
| L-Phenylalanine | 165.0 | 5.0 | 0.030303031 |
| L-Proline | 115.0 | 11.5 | 0.1 |
| L-Serine | 105.0 | 10.5 | 0.1 |
| L-Threonine | 119.0 | 3.6 | 0.0302521 |
| L-Tryptophan | 204.0 | 0.6 | 0.0029411765 |
| L-Tyrosine disodium salt dihydrate | 261.0 | 2.62 | 0.010038313 |
| L-Valine | 117.0 | 3.5 | 0.02991453 |
| Vitamins | |||
| Biotin | 244.0 | 0.024 | 9.836066E-5 |
| Choline chloride | 140.0 | 0.7 | 0.005 |
| D-Calcium pantothenate | 477.0 | 0.7 | 0.0014675052 |
| Folic Acid | 441.0 | 1.3 | 0.0029478457 |
| Niacinamide | 122.0 | 0.6 | 0.004918033 |
| Pyridoxine hydrochloride | 206.0 | 0.2 | 9.708738E-4 |
| Riboflavin | 376.0 | 0.4 | 0.0010638298 |
| Thiamine hydrochloride | 337.0 | 1.0 | 0.002967359 |
| Vitamin B12 | 1355.0 | 1.4 | 0.0010332103 |
| i-Inositol | 180.0 | 0.5 | 0.0027777778 |
| Inorganic Salts | |||
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 33.29 | 0.29990992 |
| Cupric sulfate (CuSO4-5H2O) | 250.0 | 0.0025 | 1.0E-5 |
| Ferric sulfate (FeSO4-7H2O) | 278.0 | 0.834 | 0.003 |
| Magnesium Sulfate (MgSO4) (anhyd.) | 120.0 | 74.62 | 0.6218334 |
| Potassium Chloride (KCl) | 75.0 | 285.0 | 3.8 |
| Potassium Phosphate monobasic (KH2PO4) | 136.0 | 83.0 | 0.6102941 |
| Sodium Bicarbonate (NaHCO3) | 84.0 | 1200.0 | 14.285714 |
| Sodium Chloride (NaCl) | 58.0 | 6900.0 | 118.965515 |
| Sodium Phosphate dibasic (Na2HPO4) anhydrous | 142.0 | 153.7 | 1.0823944 |
| Zinc sulfate (ZnSO4-7H2O) | 288.0 | 0.0288 | 1.0E-4 |
| Other Components | |||
| D-Glucose (Dextrose) | 180.0 | 1100.0 | 6.111111 |
| HEPES | 238.0 | 5958.0 | 25.033613 |
| Hypoxanthine Na | 159.0 | 4.7 | 0.029559746 |
| Lipoic Acid | 206.0 | 0.2 | 9.708738E-4 |
| Phenol Red | 376.4 | 1.2 | 0.0031880978 |
| Sodium Pyruvate | 110.0 | 110.0 | 1.0 |
| Thymidine | 242.0 | 0.7 | 0.002892562 |
11550 - Ham's F-10 Nutrient Mix
Cat No:
11550043
31550 - Ham's F-10 Nutrient Mix
Cat No:
31550015
,
31550023
,
31550031
,
31550049
22390 - Ham's F-10 Nutrient Mix
Cat No:
22390017
,
22390025
,
22390058
,
22390082