Search Thermo Fisher Scientific
Grant Craig, Nicholas Lloyd, Claudia Bouman, Johannes Schwieters, Thermo Fisher Scientific Bremen, Bremen, Germany; Daniel A. Frick, Helmholtz Centre Potsdam GFZ German Research Centre for Geosciences, Potsdam, Germany
In laser ablation analysis the choice of which inductively coupled plasma mass spectrometer (ICP-MS) type to use is dependent on the application. Chronometers requiring epsilon level precision are measured on multi-collector ICP-MS; determination of the trace elemental composition of a material is more appropriate on Quadrupole based instruments.
Julian D. Wills, Grant Craig, Lothar Rottmann, Thermo Fisher Scientific, Bremen, Germany; Noel D. Casey, Lee Goldstein, Boston University School of Medicine, Boston, MA , USA; Dhinesh Asogan, Teledyne Cetac Technologies, Omaha, NE, USA
Elemental mapping in cells and tissues is an important technique for improving our understanding of the role of metals in various biological processes as well as in emerging diagnostic techniques. Through instrumental advances in laser sampling (for example fast washout laser sample cells) and the increased scan speed, sensitivity and specificity of modern ICP-MS instruments, the scale of the spatial measurement of metals is ever decreasing. In this poster, the LA-ICP-MS workflow for spatial elemental analysis in a range of biological samples will be described.
Meet the ever-expanding application demands of earth sciences, nuclear sciences and other fields requiring high-precision isotope ratio measurements with the Thermo Scientific™ Neptune Plus™ high resolution multicollector ICP-MS system. The Neptune Plus system combines field-proven technology with the latest innovations.
This powerful double-focusing mass spectrometer, with high mass resolution, variable multicollectors, and multi ion counting capability offers ground-breaking sensitivity, high dynamic ranges, unsurpassed linearity, and robust stability.
The high performance, double focusing magnetic sector field Thermo Scientific™ Element 2 ™ ICP-MS is optimized for ease-of-use, stability and productivity, providing access to a powerful technique for solving a specific problem with excellent sensitivity and signal-to-noise ratio.
The Thermo Scientific™ Element XR ™ ICP-MS combines a dual mode SEM with a Faraday detector, enabling the linear dynamic range to be increased by an additional three orders of magnitude relative to the Element 2 ICP-MS.
Expand your analytical capabilities with this complete trace elemental analysis solution for your high-throughput lab. User-inspired hardware and software combine in the Thermo Scientific™ iCAP™ RQ ICP-MS to deliver maximized productivity and robustness. Simplicity and ease-of-use work in concert to streamline work-flows and achieve 'right-first-time' results; essential to all busy labs.
This innovative single quadrupole (SQ) ICP-MS is the ideal trace elemental analyzer for a wide range of sample types and industries; from 24/7 routine analysis to advanced applications in research.