Chemicals for Life Sciences Workflows
On this page:
Simplify Life Science Research: Find Chemicals for Every Step
Welcome to your chemical destination for life science research, where you can seamlessly shop for chemicals tailored to each step of your workflow. From cell culture to protein biology and molecular biology, our curated selection ensures you have the right products at every stage of your scientific journey. Simplify your research process and find exactly what you need to drive your discoveries forward with ease.
Cell biology
Our extensive range of high-quality reagents, buffers, and media are designed to support your research and innovation. Whether you're exploring cellular mechanisms, developing new therapies, or conducting advanced experiments, our products ensure precision and reliability. Discover how our chemicals can elevate your research to new heights. Explore our catalog today and unlock the potential of your cell biology studies.
Cell culture
Step 1: Seed and incubate cells, monitor growth, and scale up cultures
The first step in the cell culture workflow involves the growth of cells. This begins with cell seeding, where cells are placed into appropriate culture vessels such as petri dishes, flasks, or multi-well plates. Ensuring the correct cell density is crucial to promote optimal growth. Once seeded, the cells are incubated in an environment with controlled temperature, humidity, and CO2 levels to mimic physiological conditions. Regular monitoring of the cells under a microscope is essential to observe their growth, morphology, and confluence. Adjustments to the culture conditions are made as necessary to maintain healthy growth. As the cells reach confluence, they are sub-cultured or passaged into new vessels to increase cell quantity and maintain their health.
Step 2: Assess cell viability and density
The second step focuses on analyzing the cells. This includes assessing cell viability and density using techniques such as trypan blue exclusion or automated cell counters. Functional assays are performed to evaluate cellular functions, including proliferation assays (e.g., MTT, BrdU), apoptosis assays, or metabolic activity assays. Additionally, microscopy techniques (e.g., phase-contrast, fluorescence) are used to image cells and document their morphology, tracking changes over time and in response to treatments.
Step 3: : Harvest and process cells
The final step involves processing the cells for downstream applications. Depending on the cell type, appropriate methods are used to detach and collect cells from the culture vessels, such as trypsinization for adherent cells or centrifugation for suspension cells. The cells are then prepared for further experimental use, which may include protein extraction, RNA isolation, or flow cytometry. This preparation might involve washing, lysing, or fixing the cells. 
Protein biology
We offer a comprehensive array of high-quality reagents and tools designed to streamline your protein research, from expression and purification to analysis. Whether you're investigating protein structure, function, or interactions, our products are meticulously crafted to support your scientific journey. Explore our diverse offerings and take your protein biology research to the next level.
Step 1: Identify and characterize bioactive compounds
The process begins with the identification and characterization of bioactive molecules. These molecules can be small compounds, peptides, or other entities that interact with target proteins to modulate their function. Screening techniques such as high-throughput screening (HTS), computational modeling, and bioassays are employed to discover potential bioactive molecules. Once identified, these molecules are further characterized to understand their mechanism of action, binding affinity, and specificity towards the target protein.
Step 2: Clone and express proteins
Protein expression involves the production of the target protein in a suitable host system. The gene encoding the protein of interest is cloned into an expression vector, which is then introduced into host cells such as bacteria, yeast, insect, or mammalian cells. These cells are cultured under optimal conditions to induce the expression of the protein. The expression system and conditions are carefully optimized to achieve high yield and proper folding of the recombinant protein.
Step 3: Prepare optimal buffers
Buffers are essential for maintaining the stability and activity of proteins throughout the workflow. Different stages of protein biology require specific buffer formulations. For example, lysis buffers are used to break open cells and release proteins, while purification buffers are designed to facilitate the separation of the target protein from other cellular components. Storage buffers are optimized to preserve the protein's stability and activity over time. The pH, ionic strength, and composition of buffers are carefully adjusted to match the requirements of each step.
Step 4: Purify proteins
Protein purification is the process of isolating the target protein from a complex mixture, such as a cell lysate. This step typically involves a series of chromatography techniques, including affinity chromatography, ion exchange chromatography, and size exclusion chromatography. Each technique exploits different properties of the protein, such as its affinity for a ligand, charge, or size, to achieve separation from other proteins and contaminants. The goal is to obtain a highly pure and functional protein. 
Step 5: Analyze proteins
Protein analysis involves a range of techniques to identify and characterize the purified protein. SDS-PAGE (sodium dodecyl sulfate-polyacrylamide gel electrophoresis) is commonly used to determine the molecular weight and purity of the protein. Western blotting allows for the specific detection of the target protein using antibodies. Mass spectrometry provides precise molecular weight determination and can identify post-translational modifications. Functional assays, such as enzyme activity measurements or binding studies, confirm the biological activity of the protein.
Molecular biology
Whether you're delving into DNA sequencing, gene expression, or cloning, our products are designed to deliver unmatched accuracy and efficiency. Join the community of scientists who trust us to power their innovation and unlock the secrets of life at the molecular level. Explore our offerings today and transform your research into remarkable breakthroughs!
Step 1: Prepare optimal buffers
Buffers are essential for maintaining the stability and activity of nucleic acids and enzymes throughout the molecular biology workflow. Different stages require specific buffer formulations, such as lysis buffers for cell disruption, reaction buffers for enzymatic processes, and storage buffers for preserving nucleic acids. The composition of buffers is optimized to ensure the best conditions for each step.
Step 2: Nucleic acid and purification
Nucleic acid purification involves isolating DNA or RNA from various biological samples. Techniques such as spin column purification, magnetic bead-based purification, and organic extraction methods (e.g., phenol-chloroform) are employed to achieve high purity and yield of nucleic acids. This step is crucial for downstream applications, ensuring the nucleic acids are free from contaminants that could interfere with subsequent processes.
Step 3: Amplify DNA sequences
Endpoint PCR is a technique used to amplify specific DNA sequences, allowing for the detection and quantification of target genes. This involves the use of primers, DNA polymerase, and thermal cycling to create multiple copies of the target DNA.
Step 4: Visualize your ePCR amplicons
Nucleic acid electrophoresis is a widely used laboratory technique for separating DNA or RNA molecules based on their size. The choice of chemicals is critical at every step of this process — from the gel composition, which determines resolution, to the buffers that maintain pH and conductivity, and the dyes or stains that make nucleic acids visible. These reagents work together to ensure accurate separation, reliable results, and reproducible data, making electrophoresis a cornerstone of molecular biology workflows. Nucleic acid electrophoresis is a widely used laboratory technique for separating DNA or RNA molecules based on their size. The choice of chemicals is critical at every step of this process — from the gel composition, which determines resolution, to the buffers that maintain pH and conductivity, and the dyes or stains that make nucleic acids visible. These reagents work together to ensure accurate separation, reliable results, and reproducible data, making electrophoresis a cornerstone of molecular biology workflows.
Step 5: Cloning
Cloning is the process of inserting a DNA fragment into a vector and introducing it into host cells to replicate and express the genetic material. This involves cutting the DNA and vector with restriction enzymes, ligating the DNA fragment into the vector, and transforming the recombinant vector into competent cells. Screening and selection of positive clones are performed to identify cells that have successfully incorporated the desired DNA.
Gene expression
Quantitative PCR (qPCR), also known as real-time PCR, is an essential molecular biology technique used to amplify and accurately measure DNA or RNA. The performance of a qPCR assay depends heavily on its chemical components — including qPCR primers, nucleotides (dNTPs), thermostable DNA polymerase, fluorescent probes or dyes, and optimized reaction buffers. Each of these reagents is carefully designed to maximize sensitivity, specificity, and reproducibility, ensuring reliable detection and quantification of target genetic material. Choosing the right qPCR reagents and workflow conditions is critical for obtaining consistent, high-quality results.
For Research Use Only. Not for use in diagnostic procedures.