Thermo Scientific SamStat-30 Representative Slurry Sampling Stations and AnStat-330 Online Sampling and Elemental Analysis Stations both provide periodic composite metallurgical samples for accounting while providing online elemental analysis. Use this comparison chart to see which system is right for you.
Product Family |
Differentiating Features |
Metallurgical Composite Sampler |
Analyzer Type |
Pebble Screen |
High Froth |
Sample Take-off |
Distributor |
SamStat-30 |
Upgradable to AnStat |
Y |
N |
N |
N |
N |
N |
SamStat-30F |
Upgradable to AnStat |
Y |
N |
N |
Y |
N |
N |
SamStat-30D |
Upgradable to AnStat |
Y |
N |
N |
N |
|
Y |
SamStat-30T |
Upgradable to AnStat |
Y |
N |
N |
N |
Y |
N |
Sam-Stat30P |
Upgradable to AnStat |
Y |
N |
Y |
N |
N |
N |
SamStat-30DP |
Upgradable to AnStat |
Y |
N |
Y |
N |
N |
Y |
SamStat-30DT |
Upgradable to AnStat |
Y |
N |
N |
N |
Y |
Y |
SamStat-30PT |
Upgradable to AnStat |
Y |
N |
Y |
N |
Y |
N |
SamStat-30DPT |
Upgradable to AnStat |
Y |
N |
Y |
N |
Y |
Y |
|
|||||||
SamStat-30C |
Continuous Flow +Incremental Sampling |
N |
N |
N |
N |
N |
N |
SamStat-30CP |
Continuous Flow + Pebble Screen |
N |
N |
Y |
N |
N |
N |
SamStat-30CF |
Continuous Flow + Froth
|
N |
N |
N |
Y |
N |
N |
SamStat-30CT |
Continuous Flow + Take Off Point |
N |
N |
N |
N |
Y |
N |
SamStat-30CD |
Continuous Flow + Distributor |
N |
N |
N |
N |
N |
Y |
SamStat-30CDP |
Continuous Flow + Distributor + Pebble Screen |
N |
N |
Y |
N |
N |
Y |
SamStat-30CDT |
Continuous Flow + Distributor + Take Off Point |
N |
N |
N |
N |
Y |
Y |
SamStat-30CPT |
Continuous Flow + Pebble Screen + Take Off Point |
N |
N |
Y |
N |
Y |
N |
SamStat-30CDPT |
Continuous Flow + Distributor + Pebble Screen + Take Off Point |
N |
N |
Y |
N |
Y |
Y |
SamStat-30R
|
Primary Sampler for Particle Size Monitor + Reverse Sampling |
N |
N |
N |
N |
N |
N |
|
|||||||
GF-Samp |
Gravity Pipe Sampler |
N |
N |
N |
N |
N |
N |
PP-Samp |
Pressure Pipe Sampler |
N |
N |
N |
N |
N |
N |
X-Cut Samp |
Cross Cut Composite Sampler |
Y |
N |
N |
N |
N |
N |
Product Family |
Differentiating Features |
Metallurgical Composite Sampler |
Analyzer Type |
Pebble Screen |
High Froth |
Sample Take-off |
Distributor |
AnStat-330 |
Generation 3 |
Y |
XRF |
N |
N |
N |
N |
AnStat-330P |
Generation 3 |
Y |
XRF
|
Y |
N |
N |
N |
AnStat-330F
|
Generation 3 |
Y |
XRF
|
N |
Y |
N |
N |
AnStat-330T |
Generation 3 |
Y |
XRF
|
N |
N |
Y |
N |
AnStat-330T2
|
Generation 3 |
Y |
XRF
|
N |
N |
Y-2 |
N |
AnStat-330T3 |
Generation 3 |
Y |
XRF
|
N |
N |
Y-3 |
N |
AnStat-330D |
Generation 3 |
Y |
XRF
|
N |
N |
N |
Y |
AnStat-330PT |
Generation 3 |
Y |
XRF
|
Y |
N |
Y |
N |
AnStat-330DP |
Generation 3 |
Y |
XRF
|
Y |
N |
N |
Y |
AnStat-330DT |
Generation 3 |
Y |
XRF
|
N |
N |
Y |
Y |
AnStat-330DPT |
Generation 3 |
Y |
XRF
|
Y |
N |
Y |
Y |
Product Family |
Differentiating Features |
Process Type |
Analysis Type |
Method |
Source |
Streams |
Composite & Calibration Sampler |
AnStat-330 |
Sampler/Analyzer |
Slurry |
Elemental |
XRF |
Isotope |
1 |
Y |
MSA-330 |
Multi-stream Centralized |
Slurry
|
Elemental |
XRF |
Isotope |
3-12 |
Y |
GS-Omni |
Multi-stream Centralized |
Slurry
|
Elemental |
PGNAA |
Isotope |
2-8 |
Y |
PSM-500 |
Sample Conditioner/Analyzer |
Slurry
|
Particle Size |
Ultrasonic Absorption |
n/a |
1-3 |
Y |
CB-Omni Agile |
Single stream |
Conveyor Belt |
Elemental |
PGNAA |
Isotope nGen |
1 |
Y |