Need a different kind of vial?
We offer selection of vials and closures to match your application and autosampler requirements. These specialty vials include EPA, headspace, pre-cleaned, reaction, 4mL, sample storage and shell vials.
Featured specialty vials
Recommended for discrete water sampling under EPA 40 CFR 136 “Guidelines for Establishing Test Procedures for the Analysis of Pollutants” and EPA 40 CFR 141 “National Interim Primary Drinking Water Regulations: Control of Trihalomethanes in Drinking Water.”
Headspace Vials meet or exceed OEM specifications to ensure autosampler compatibility.
Manufactured from high quality glass, these 4mL (13mm), 15x45mm vials feature 13-425 thread finish and fit a variety of autosamplers.
Store your samples with confidence and convenience with these assembled screw vial kits.
Shell vials are an economical choice for many routine HPLC applications. These neckless vials with inserts are sold as kits including polyethylene push in caps to assure proper fit and function.
Select Chromatography Consumables and Columns for your Pharma, Biopharma, Clinical, Food & Beverage or Environmental workflow.
Access the latest version of our Chromatography Consumables Catalog to ensure that you're optimizing your applications! Our enhanced online version features a robust applications search tool, access to method development guides, application notebooks, and much more.