Make the most of small sample volumes
The base of all microvials are designed to recover the maximum amount of sample with a minimum amount of sample waste, whether using:
- One-piece microvials
- Inserts in a regular vial
- Conical microvials in a sleeve
- High throughput microplates
Our broad variety of Thermo Scientific microvials address a broad range of sample requirements, applications, and autosampler compatibility.
One-piece microvials
One-piece microvials have the insert conveniently built into the vial and require less handling of vial components compared to using separate inserts. Click on the options below to see Thermo Scientific one-piece microvials size and shape of your choice. They are available in a variety of sizes and materials to satisfy your sample requirements and autosampler compatibility.
8mm (8-425) Screw Thread Microvials | ||
---|---|---|
plastic | ||
C4013-11 | 8mm (8-425) Polypropylene Screw 300µL* Microvial 12x32 Inner Conical Shape 100/PK | |
C4013-13 | 8mm (8-425) Polypropylene Screw 675µL* Microvial 12x32 Inner Conical Shape 1000/PK | |
03-PPSV | 8mm (8-425) Polypropylene Screw 250µL* Microvial 12x32 Inner Conical Shape 500/PK | |
Clear glass | ||
1.1-STVG | 8mm (8-425) GOLD Grade Clear Glass Screw 1100μL* Microvial 12x32 Tapered Conical 500/PK |
* Useable volume for Microvial
9mm Screw Thread Microvials | |
---|---|
Plastic | |
03-PPSVW | 9mm Polypropylene Screw 200µL* Microvial 12x32 Inner Conical Shape 500/PK |
C4000-11 | 9mm Target DP Polypropylene Screw 300µL* Microvial 12x32 Inner Conical Shape 100/PK |
Clear glass | |
02-FISVG | 9mm GOLD Grade Clear Glass Screw 180µL* Microvial 12x32 Conical Insert 500/PK |
C4000-LV3W | 9mm Target DP Clear Glass Screw 240μL* Microvial 12x32 Conical Insert 100/PK |
03-FISV | 9mm Clear Glass Screw 250µL* Microvial 12x32 Conical Insert 500/PK |
60180-507 | 9mm Clear Glass Screw w/fused 250µL* Microvial 12x32 Conical Insert 100Pk |
C4000-LV1 | 9mm Target DP Clear Glass Screw 350µL* Microvial 12x32 Conical Insert 100/PK |
C4000-LV1W | 9mm Target DP Clear Glass Screw 350µL* Microvial 12x32 with patch Conical Insert 100/PK |
C4000-V1 | 9mm Target DP Clear Glass Screw 1000μL* MICRO-V Microvial with 150µL reservoir, 12X32mm High Recovery 100/PK |
1.2-UHRSV | 9mm Clear Screw 1000µL*Microvial, Ultra High Recovery with 10µL Reservoir, 12x32 100/PK |
C4000-9TR | 9mm Target DP Clear Glass Screw 1200 µL* Microvial, Total Recovery with 10µL Reservoir, 12x32 100/PK |
1.5-HRSV | 9mm Clear Glass Screw 1300µL* Microvial, 12x32 High Recovery with 30µL Reservoir |
C4000-9 | 9mm Target DP Clear Glass Screw 1300µL*Microvial with 30µL Reservoir 12x32 High Recovery 100/PK |
C4000-S9 | 9mm Target DP Silanized Clear Glass Screw 1300μL* Microvial with 30μL Reservoir, High Recovery 12x32 100/PK |
Amber glass | |
03-FISV(A) | 9mm Amber Glass Screw 250µL* Microvial 12x32 Conical Insert 500/PK |
C4000-LV2 | 9mm Target DP Amber Glass Screw 350µL* Microvial 12x32 Conical Insert 100/PK |
C4000-LV2W | 9mm Target DP Amber Glass Screw 350µL* Microvial 12x32 with patch Conical Insert 100/PK |
C4000-V2 | 9mm Target DP Amber Glass Screw 1000μL* MICRO-V Microvial with 150µL reservoir, 12X32mm High Recovery 100/PK |
C4000-9A | 9mm Target DP Amber Glass Screw 1300µL* Microvial with 30µL Reservoir 12x32 High Recovery 100/PK |
* Useable volume for Microvial
9mm wide opening screw top microvial convenience kits with AVCS caps** | ||||
---|---|---|---|---|
Convenience Kit | Includes Vial | Includes Septa | Includes AVCS Cap | Product Size |
C5000-87 | 9mm Target DP Polypropylene 300µL* Microvial C4000-11 | Blue Red PTFE/White Silicone | AVCS Blue Cap C5000-54B | Pack of 100 |
C5000-LV95 | 9mm Target DP Clear Glass Screw 350µL* Microvial 12x32 Conical Insert C4000-LV1 | PTFE/White Silicone, Pre-slit | AVCS Blue Cap C5000-54B | Pack of 100 |
C5000-97 | 9mm Target DP Polypropylene 300µL* Microvial C4000-11 | Blue PTFE/White Silicone, Pre-slit | AVCS Blue Cap C5000-54B | Pack of 100 |
* Useable volume for Microvial
** AVCS caps designed to eliminate failures by preventing push-through of the septa and overtightening of the cap. We offer the following microvial kits with AVCS caps.
9mm Certified Microvial Kits** | ||||
---|---|---|---|---|
Convenience Kit | Includes Vial | Includes Septa | Includes AVCS Cap | Product Size |
CERT5000-69LV | 9mm Target DP Clear Glass Screw 350µL* Microvial 12x32 with patch Conical Insert C4000-LV1W | Bonded Red PTFE/White Silicone | Black | Pack of 100 |
CERT5000-72LV | 9mm Target DP Clear Glass Screw 350µL* Microvial 12x32 with patch Conical Insert C4000-LV1W | Bonded Red PTFE/White Silicone | Black | Pack of 100 |
CERT5000-993 | 9mm Target DP Clear Glass Screw 1200 µL* Microvial, Total Recovery with 10 µL Reservoir, 12x32 C4000-9TR | Bonded Clear PTFE/Clear Silicone, Pre-slit | Gray | Pack of 100 |
CERT5000-79 | 9mm Target DP Clear Glass Screw 1300µL* Microvial with 30µL Reservoir 12x32 High Recovery Microvial C4000-9 | Bonded Clear PTFE/Clear Silicone | Gray | Pack of 100 |
* Useable volume for Microvial
** Certified kits are fully lot-tested including HPLC and GC analysis for 15 critical parameters
9mm MS-Certified Microvial Kits** | ||||
---|---|---|---|---|
Convenience Kit | Includes Vial | Includes Septa | Includes AVCS Cap | Product Size |
MSCERT5000-30LVW | 9mm Target DP Clear Glass Screw 240 μL* Microvial 12x32 Conical Insert C4000-LV3W | Bonded Clear PTFE/Clear Silicone | Blue | Pack of 100 |
MSCERT5000-31LVW | 9mm Target DP Clear Glass Screw 350 µL* Microvial 12x32 with patch Conical Insert C4000-LV1W | Bonded Clear PTFE/Clear Silicone | Blue | Pack of 100 |
MSCERT5000-32 | 9mm Target DP Clear Glass Screw 1300µL* Microvial with 30µL Reservoir 12x32 High Recovery C4000-9 | Bonded Clear PTFE/Clear Silicone | Blue | Pack of 100 |
MSCERT5000-33TR | 9mm Target DP Clear Glass Screw 1200 µL* Microvial, Total Recovery with 10 µL Reservoir, 12x32 C4000-9TR | Bonded Clear PTFE/Clear Silicone | Blue | Pack of 100 |
MSCERT5000-36LVW | 9mm Target DP Clear Glass Screw 240 μL* Microvial 12x32 Conical Insert C4000-LV3W | Bonded Clear PTFE/Clear Silicone, Pre-slit | Gray | Pack of 100 |
MSCERT5000-37LVW | 9mm Target DP Clear Glass Screw 350 µL* Microvial 12x32 with patch Conical Insert C4000-LV1W | Bonded Clear PTFE/Clear Silicone, Pre-slit | Gray | Pack of 100 |
MSCERT5000-38 | 9mm Target DP Clear Glass Screw 1300 µL* Microvial with 30µL Reservoir 12x32 High Recovery C4000-9 | Bonded Clear PTFE/Clear Silicone, Pre-slit | Gray | Pack of 100 |
MSCERT5000-39TR | 9mm Target DP Clear Glass Screw 1200 µL* Microvial, Total Recovery with 10µL Reservoir, 12x32 C4000-9TR | Bonded Clear PTFE/Clear Silicone, Pre-slit | Gray | Pack of 100 |
* Useable volume for Microvial
** MS Certified kits are the first low particle, low background chromatography vials and the only vials pre-cleaned to provide unmatched consistency
10mm (10-425) Screw Thread Microvials | |
---|---|
C4010-11 | 10mm (10-425) Polypropylene Screw 400μL* Microvial 12x32 Inner Conical Shape 100/PK |
C4010-14 | 10mm (10-425) Polypropylene Screw 550μL* Microvial 12x32 Inner Conical Shape 100/PK |
C4010-LV1 | 10mm (10-425) Clear Glass Screw 350μL* Microvial 12x32 Conical Insert 100/PK |
C4010-LV2 | 10mm (10-425) Amber Glass Screw 350μL* Microvial 12x32 Conical Insert 100/PK |
C4010-V1 | 10mm (10-425) Clear Glass Screw 1100μL* Microvial 12x32 High Recovery 100/PK |
* Useable volume for Microvial
13mm (13-425) 4mL Screw Thread Microvials | |
---|---|
C4015-9 | 13mm (13-425) Glass Screw 3500μL* Microvial 15x45 High Recovery 100/PK |
C4015-14 | 13mm (13-425) Polypropylene Screw 2000μL* Microvial 15x45 Tapered Base High Recovery 100/PK |
3.5-HRSV | 13mm (13-425) Clear Glass Screw w/fused 3000µL* Microvial 15.25x46 High Recovery 250/PK |
5-SV | 13mm (13-425) Clear Glass Screw w/fused 4500µL* Microvial 13x65 Round Base 125/PK |
* Useable volume for Microvial
8mm Crimp Top Vials for Microsampling | |
---|---|
8mm Clear glass crimp top microvials | |
01-CVG | Chromacol Inert Vial 8mm GOLD Grade Clear Glass Sci-Vi Crimp 80µL* Microvial 6x32 Round Bottom 500/PK |
02-CTVG | Chromacol Inert Vial 8mm GOLD Grade Clear Glass Sci-Vi Crimp 200µL* Microvial 6x32 Tapered 500/PK |
03-CVG | Chromacol Inert Vial 8mm GOLD Grade Clear Glass Sci-Vi Crimp 250µL* Microvial 6x32 Round Base 500/PK |
C4008-632C | 8mm Clear Glass Crimp 200µL* Microvial, 6x32 Conical Base 100/PK |
C4008-632R | 8mm Clear Glass Crimp 250µL* Microvial, 6x32mm Round Base 100/PK |
C4008-741 | 8mm Clear Glass Crimp 650µL* Microvial, 7x40mm Flat Base 100/PK |
07-CPV | 8mm Clear Glass Crimp 450µL* Microvial 7x32 Conical Base 500/PK |
08-CPV | 8mm Clear Glass Crimp 650µL* Microvial 7x40 Flat Base 500/PK |
C4008-1 | 8mm Clear Glass Crimp 800µL* Microvial, 8x30 Flat Base 200/PK |
08-CV | 8mm Clear Glass Crimp 800µL* Microvial 8x30 Flat Base 500/PK |
1-CWV | 8mm Clear Glass Crimp 1000µL* Microvial 8x40 Tapered 500/Pack |
1.2-CWV | 8mm Clear Glass Crimp 1200µL* Microvial 8x40 Flat Base 500/PK |
8mm amber glass crimp top microvials | |
02-CTV(A) | 8mm Amber Glass Sci-Vi Crimp 200µL* Microvial 6x32 Conical Base 500/Pack |
C4008-740 | 8mm Amber Glass Crimp 450µL* Microvial, 7x40mm Conical Base 100/PK |
05-CTV(A) | 8mm Amber Glass Crimp 450µL* Microvial 7x30 Tapered 500/PK |
07-CPV(A) | 8mm Amber Glass Crimp 450µL* Microvial 7x40 Tapered 500/PK |
06-CTV(A) | 8mm Amber Glass Crimp 550µL* Microvial 7x32 Tapered 500/PK |
08-CRV(A) | 8mm Amber Glass Crimp 600µL* Microvial 7x32 Round Base 500/PK |
C4008-742 | 8mm Amber Glass Crimp 650µL* Microvial, 7x40mm Flat Base 100/PK |
* Useable volume for microvial
11mm Crimp Microvials | |
---|---|
Clear glass | |
02-FIVG | Chromacol Gold Inert Vial 11mm Clear GOLD Grade Glass 180µL* Microvial 12x32 Conical Insert 500/PK |
1.1-CTVG | Chromacol Gold Inert Vial 11mm GOLD Grade Clear Glass Crimp 1100µL* Microvial 12X32 Conical Base 500/PK |
03-FIV | 11mm Clear Glass Crimp 250µL* Microvial 12x32 Conical Insert 500/PK |
C4011-LV1W | 11mm Clear Glass Crimp 350µL* Microvial 12x32 Conical Insert 100/PK |
09-FIV | 11mm Clear Glass Crimp 830µL* Microvial 12x32 Conical Insert 500/PK |
09-CTV | 11mm Clear Glass Crimp 850µL* Microvial 10x32 Conical Base 500/PK |
C4011-V1 | 11mm Clear Glass Crimp 1100µL* Microvial 12x32 High Recovery Micro-V 100/PK |
1.5-HRCV | 11mm Clear Glass Crimp 1300µL* Microvial 12x32 High Recovery 100/PK |
2-CRV | 11mm Clear Glass Crimp 1500µL* Microvial 12x32 Round Base 500/PK |
Amber glass | |
03-FIV(A) | 11mm Amber Glass Crimp 250µL* Microvial 12x32 Conical Insert 500/PK |
C4011-LV2W | 11mm Amber Glass Crimp 350µL* Microvial 12x32 Conical Insert 100/PK |
1.1-CTV(A) | 11mm Amber Glass Crimp 1100µL* Microvial 12x32 Conical Base 500/PK |
11mm Crimp/SNAP and Snap-IT Microvials | |
Plastic | |
C4011-13 | 11mm Polypropylene Crimp/Snap 250µL* Microvial 12x32 Inner Conical Shape 100/PK |
C4011-16 | 11mm Polypropylene Crimp/Snap 400µL* Microvial 12x32 Inner Conical Shape 100/PK |
C4011-11 | 11mm Polypropylene Crimp/Snap 600µL* Microvial 12x32 Inner Conical Shape 100/PK |
C4011-14 | 11mm Polypropylene Crimp/Snap 800µL* Microvial 12x32 Inner Flat Shape 100/PK |
C4011-15 | 11mm Polypropylene Snap-It 650µL* Microvial 12x32 High Recovery 1000/PK |
C4011-24 | 11mm TPX TARGET Snap-It 7500µL* Microvial 12x32 High Recovery 100/PK |
Clear Glass | |
03-FIRV | 11mm Clear Glass Snap-Cap 250µL* Microvial 12x32 Conical Insert 500/PK |
C4012-15 | 11mm Clear Glass Crimp/Snap 350µL* Microvial TPX Vial with Glastic Conical Glass Insert, 12x32, 100/PK |
C4011-LV1 | 11mm TARGET Clear Glass Snap 350µL* Microvial 12x32 Conical Insert 100/PK |
1.2-UHRRV | 11mm Clear Glass Snap-Cap, 1000µL* Microvial 12x32 Mandrel Base Ultra High Recovery with 10µL Reservoir 100/PK |
C4011-9TR | 11mm Clear Glass Total Recovery Snap-It 1200µL* Microvial with 10µL Reservoir12x32 Deep Well Base 100/PK |
C4011-4 | 11mm Clear Glass Snap-It 1300µL* Microvial 12x32 High Recovery 100/PK |
1.5-HRRV | 11mm Clear Glass Snap Cap 1300µL* Microvial 12x32 High Recovery 100/PK |
Silanized clear glass | |
C4011-S4 | 11mm Silanized Clear Glass Snap-It 1100µL* Microvial 12x32 High Recovery 100/PK |
1.5-HRRV(S) | 11mm Silanized Clear Glass Snap Cap 1300µL* Microvial 12x32 High Recovery 100/PK |
Amber glass | |
03-FIRV(A) | 11mm Amber Glass Snap-Cap 250µL* Microvial 12x32 Conical Insert 500/PK |
C4011-LV2 | 11mm Amber Glass Snap 350µL* Microvial 12x32 Conical Insert 100ist. |
* Useable volume for Microvial
Microvials can be sealed, but often need an adapter to run in the autosampler. In case they have a conical bottom, they cannot stand by themselves.
Inserts for regular vials
Plastic, glass, or silanized glass inserts are placed inside of 2mL crimp, snap, or screw thread vials. The diameter of the insert you choose is dependent on the size of the vial opening.
- Inserts for 8 mm small ID vials (5 mm ID inserts)
- Inserts for 9 mm wide ID vials (6 mm ID inserts)
- Inserts for 11 mm wide ID vials (6 mm ID inserts)
- Inserts for 13 mm (4 mL) vials
WebSeal 96- and 384-well microplates & mats
- WebSeal Plate+ 384-Well Glass-Coated Microplates
- WebSeal Plate+ 96-Well Glass-Coated Microplates
- WebSeal 384-Well Non-Coated Plastic Microplates
- WebSeal 96-Well Non-Coated Plastic Microplates
- WebSeal 96-Well Plate Glass Inserted Kits with Sealing Mats
- WebSeal 96-Well Plate Glass Inserted Kits with Individual Closures