Search Thermo Fisher Scientific
Our complete testing workflows help you to ensure safe, clean, and compliant water. From peptones to plates and filters, our high-quality media are compliant and certified according to ISO requirements for performance filter combination testing. We also have full manufacturing process traceability from raw materials to final products, ensuring rigorous quality and complete confidence.
Conveniently prepare water samples when isolating and enumerating Legionella species according to EN ISO 11731 water quality testing specifications using Thermo Scientific Legionella Acid Buffer Solution. The convenient high-quality, transparent PET bottle has a leak-proof screw top made of PET and is lighter and more shatter-resistant than glass, making it better for handling in the laboratory as well as shipping.
To support your laboratory’s water testing processes, we have added Thermo Scientific Culti-Loops Quality Control Legionella anisa NCTC 11974 (WDCM 00106) to our quality control testing organism range. Culti-Loops Quality Control organisms enable quick and safe preparation of cultures for QC testing with ready-to-use, disposable inoculation loops containing gel-stabilised micro-organisms.
Legionella anisa is the second most often isolated Legionella species from water samples, after Legionella pneumophila.
We guarantee the performance of your prepared plate media products throughout their entire shelf life. We conduct simulation studies according to SOP BT-SOP-6958 procedure (ISO) to assess product performance following room temperature transportation. Use the transport data captured by the Data Logger USB to ensure that the validated room temperature storage parameters, or shelf-life report with transportation simulation, have not been exceeded.
Method |
Media | Product | Format | Product Code |
---|---|---|---|---|
Pour plate | Yeast Extract Agar | Thermo Scientific Water Plate Count Agar (Dehydrated) | 500 g Dehydrated culture media |
CM1012B |
Thermo Scientific Water Plate Count Prepared Agar, Bottled | 10 x 100 mL bottles Prepared media |
BO0055M | ||
10 x 200 mL bottles Prepared media |
BO0055R |
Method | Media | Product | Format | Product code |
---|---|---|---|---|
Membrane filtration | CCA Agar | Thermo Scientific Chromogenic Coliform Agar | 500 g Dehydrated culture media |
CM1205B |
10 x 90 mm plates Prepared media |
PO5318A | |||
10 x 55 mm plates Prepared media |
PO5428J | |||
Oxidase test | Oxidase Reagent |
Thermo Scientific BactiDrop Oxidase Reagent |
50 vials Dehydrated culture media |
R21540 |
10 x 90 mm plates Prepared media |
PO5321A | |||
Thermo Scientific Microbact Oxidase Strips | 50 strips Dehydrated culture media |
MB0266A |
Method | Media | Product | Format | Product code |
---|---|---|---|---|
Membrane filtration | Slanetz Bartley Agar | Thermo Scientific Slanetz Bartley Agar (Enterococcus Agar) | 500 g Dehydrated culture media |
CM0377B |
10 x 90 mm plates Prepared media |
PO5018A | |||
10 x 55 mm plates Prepared media | PO5410J* | |||
PO5423J* | ||||
Confirmation | Bile Aesculin Azide Agar | Thermo Scientific Bile Esculin Azide Agar (Enterococcus selective medium)1 | 10 x 90 mm plates Prepared media |
PO5062A |
Thermo Scientific Kanamycin Aesculin Azide Agar Base2 | 500 g Dehydrated culture media |
CM0591B | ||
Thermo Scientific Kanamycin Aesculin Azide Selective Medium | 10 x 90 mm plates Prepared media |
PO5059A |
* Minimum order quantity required.
1. This medium is slightly different to the medium formulation according to ISO 7899-2:2000. The medium has additional sodium citrate and 20 g/L ox bile instead of 10 g/L and 0.55 g/L sodium azide instead of 0.15 g/L.
2. This medium is slightly different to the medium formulation according to ISO 7899-2:2000. It contains sodium citrate instead of ox bile and Kanamycin.
Method | Media | Product | Format | Product code |
---|---|---|---|---|
Membrane filtration | Pseudomonas Selective Agar / CN Agar | Thermo Scientific Pseudomonas C-N Selective Agar (Cetrimide Agar) | 10 x 90 mm plates Prepared media |
PO5076A |
10 x 55 mm plates Prepared media |
PO5413J* | |||
Thermo Scientific Pseudomonas Agar Base | 500 g Dehydrated culture media |
CM0559B | ||
Selective supplement | Thermo Scientific Oxoid C-N Supplement | 10 vials Each vial for 500mL medium |
SR0102E | |
Confirmation | ||||
Fluorescence | King´s B Media | |||
Hydrolysis of acetamide to ammonia | Acetamide Solution Nessler Reagent | |||
Oxidase Test | Nutrient Agar | Thermo Scientific Nutrient Agar | 500 g Dehydrated culture media |
CM0003B |
Oxidase Reagent | Thermo Scientific BactiDrop Oxidase Reagent | 50 vials Dehydrated culture media |
R21540 | |
Thermo Scientific Microbact Oxidase Strips | 50 strips Dehydrated culture media |
MB0266A |
* Minimum order quantity required.
Method | Media | Product | Format | Product code |
---|---|---|---|---|
Membrane filtration | TSC Agar | Thermo Scientific Perfringens TSC Selective Agar | 10 x 90 mm plates Prepared media |
PO5315A |
Thermo Scientific Perfringens TSC Agar Base + | 500 g Dehydrated culture media |
CM0587B | ||
50 tubes Prepared media |
TV5204G | |||
Thermo Scientific Perfringens TSC Selective Supplement | 10 tubes Dehydrated culture media |
SR0088E | ||
10 x 100 mL bottles Prepared media |
BO0634M | |||
Anaerobic atmosphere | Thermo Scientific AnaeroGen System | 10 sachets |
AN0010C | |
Thermo Scientific AnaeroJar 2.5 L Anaerobic System | 2.5 L jar | AG0025A | ||
Thermo Scientific AnaeroBox 2.5 L Rectangular Anaerobic System | 1 unit |
AB0025A |
Method | Media | Product | Format | Product code |
---|---|---|---|---|
Sample preparation | Acid Buffer | Legionella Acid Buffer Solution | 4 x 1 L bottles Prepared media |
EB1375W |
Direct and membrane filtration | Legionella GVPC Agar | Legionella GVPC Selective Agar | 10 x 90 mm plates Prepared media |
PO5074A |
10 x 90 mm plates Prepared media |
PO0245A | |||
Legionella CYE Agar Base + | 500 g Dehydrated culture media |
CM1203B | ||
Legionella BCYE Supplement + | 10 tubes (500 mL) Dehydrated culture media |
SR0251C | ||
Legionella GVPC Selective Supplement | 10 tubes (500 mL) Dehydrated culture media |
SR0252E | ||
Legionella GVPC Agar with Antibiotics | Legionella BCYE Agar with Antibiotics | 10 x 90 mm plates Prepared media |
PO5325A | |
Legionella BCYE Agar | Legionella BCYE Agar | 10 x 90 mm plates Prepared media |
PO5072A | |
Legionella CYE Agar Base + | 500 g Dehydrated culture media |
CM1203B | ||
Legionella BCYE Supplement | 10 tubes (500 mL) Dehydrated culture media |
SR0251C | ||
Legionella MWY Agar | Legionella MWY Selective Agar | 10 x 90 mm plates Prepared media |
PO5071A | |
Legionella CYE Agar Base + | 500 g Dehydrated culture media |
CM1203B | ||
Legionella BCYE Supplement + | 10 tubes (500 mL) Dehydrated culture media |
SR0251C | ||
Legionella MWY Selective Supplement | 10 tubes (500 mL) Dehydrated culture media |
SR0255B | ||
Subculture | Legionella BCYE Agar without Cysteine | Legionella BCYE Agar without Cysteine | 10 x 90 mm plates Prepared media |
PO5028A |
10 x 90 mm plates Prepared media |
PO0255A* | |||
Legionella CYE Agar Base + | 500 g Dehydrated culture media |
CM1203B | ||
Legionella BCYE Supplement without Cysteine | 10 tubes (100 mL) Dehydrated culture media |
SR0253A | ||
Legionella BCYE Agar | Legionella BCYE Agar | 10 x 90 mm plates Prepared media |
PO5072A | |
Legionella CYE Agar Base + | 500 g Dehydrated culture media |
CM1203B | ||
Legionella BCYE Supplement | 10 tubes (500 mL) Dehydrated culture media |
SR0251C |
* Minimum order quantity required.
Product | Format | Order code |
---|---|---|
qualyfast Reagents and PCR Kits | ||
qualyfast Free DNA Removal | 200 reactions | D401009 |
qualyfast DNA Extraction Kit I Clear Water | 24 reactions | D101001 |
48 reactions | D201001 | |
96 reactions | D301001 | |
qualyfast DNA Extraction Kit II Dirty Water | 48 reactions | D211001 |
96 reactions | D311001 | |
qualyfast Legionella PCR Assay | 24 reactions (3 x 8-tube strips) |
A101013 |
48 reactions (6 x 8-tube strips) |
A201013 | |
96 reactions (12 x 8-tube strips) |
A301013 | |
qualyfast Legionella Quantification Standards | 8 tests (4 standards each in an 8-tube strip) |
A501013 |
QuantStudio 5 Food Safety PCR System | ||
Applied Biosystems QuantStudio 5 Food Safety Real-Time PCR System, 0.1 mL Block, Laptop | System | A36328 |
Applied Biosystems QuantStudio 5 Food Safety Real-Time PCR System, 0.1 mL Block, Desktop | System | A36320 |
Service & Support Packages | ||
Applied Biosystems Assurance Service Plan additional 1 year warranty cover with instrument loan |
1 year cover | SCQS5FAST |
Product | Size of Quantity | Catalog Number |
---|---|---|
Nalgene Single Use Analytical Filter Funnels | ||
Sterile Analytical Filter Units, 100 mL, 47 mm, 0.45 μm, white/black | 50 pieces | 145-0045 |
Sterile Analytical Filter Units, 250 mL, 47 mm, 0.45 μm, white/black | 50 pieces | 145-2045 |
Sterile Analytical Filter Units, 100 mL, 47 mm, 0.45 μm, grey/black | 50 pieces | 147-0045 |
Sterile Test Filter Funnel, 250 mL, 47 mm, 0.45 μm, grey/black | 50 pieces | 147-2045 |
Nalgene Accessories | ||
Vacuum Filter Funnel Manifold | 1 piece | DS0345-0001 |
Filter Stopper, non-sterile, no. 8 rubber | 1 piece | DS0396-0080 |
Filter Forceps, bent tip | 1 piece | DS0399-0001 |
Filter Forceps, straight tip | 1 piece | DS0399-0002 |
Filter Funnel Adapter, non-sterile | 25 pieces | DS0397-0010 |
Vacuum Gasket, non-sterile thermoplastic elastomer | 6 pieces | DS0395-0708 |
Nalgene Reusable Analytical Filters and Accessories | ||
Filter Funnels with Clamp, 250 mL | 1 piece | DS0315-0047 |
Filter Holders with Receiver, 500 mL, 500 mL | 4 pieces | 300-4000 |
Filter Holders with Receiver, 250 mL, 250 mL | 4 pieces | 300-4050 |
Filter Holders with Receiver, 500 mL, 1000 mL | 4 pieces | 300-4100PK |
Filter Holders with Funnel, 250 mL | 1 piece | DS0310-4000 |
Filter Holders with Funnel, 500 mL | 1 piece | DS0310-4050 |
Reusable Bottle Top Filters, 250 mL, 45 mm | 1 piece | DS0320-2545 |
Reusable Bottle Top Filters, 500 mL, 33 mm | 1 piece | DS0320-5033 |
Reusable Bottle Top Filters, 500 mL, 45 mm | 1 piece | DS0320-5045 |
Product | Sample numbers | Catalog Number |
---|---|---|
Escherichia coli QC Organisms for testing of CCA Agar | ||
Escherichia coli ATCC® 8739* | WDCM 00012 | R4607085 |
Escherichia coli ATCC® 25922* | WDCM 00013 | R4607050 |
Klebsiella aerogenes ATCC® 13048* | WDCM 00175 | R4607080 |
Pseudomonas aeruginosa ATCC® 27853* | WDCM 00025 | R4607060 |
Pseudomonas paraeruginosa ATCC® 9027* | WDCM 00026 | R4605210 |
Enterococcus faecalis ATCC® 19433* | WDCM 00009 | R4601990 |
Enterococcus faecalis ATCC® 29212* | WDCM 00087 | R4607030 |
Enterococci QC Organisms for testing of Slanetz and Bartley Agar | ||
Escherichia coli ATCC® 8739* | WDCM 00012 | R4607085 |
Escherichia coli ATCC® 25922* | WDCM 00013 | R4607050 |
Staphylococcus aureus subsp. aureus ATCC® 6538* | WDCM 00032 | R4607016 |
Staphylococcus aureus subsp. aureus ATCC® 25923* | WDCM 00034 | R4607010 |
Enterococcus faecalis ATCC® 29212* | WDCM 00087 | R4607030 |
Enterococcus faecalis ATCC® 19433* | WDCM 00009 | R4601990 |
Pseudomonas aeruginosa QC Organisms for testing of Pseudomonas CN Agar | ||
Pseudomonas aeruginosa ATCC® 27853* | WDCM 00025 | R4607060 |
Pseudomonas paraeruginosa ATCC® 9027* | WDCM 00026 | R4605210 |
Pseudomonas aeruginosa ATCC® 10145* | WDCM 00024 | R4607065 |
Escherichia coli ATCC® 8739* | WDCM 00012 | R4607085 |
Escherichia coli ATCC® 25922* | WDCM 00013 | R4607050 |
Enterococcus faecalis ATCC® 19433* | WDCM 00009 | R4601990 |
Enterococcus faecalis ATCC® 29212* | WDCM 00087 | R4607030 |
Clostridium perfringens QC Organisms for testing of TSC Agar | ||
Bacillus spizizenii ATCC 6633 | WDCM 00003 | R4601221 |
QC organisms for testing GCVP Agar (Legionella method) | ||
Legionella pneumophila ATCC® 33152* | WDCM 00107 | R4603950 |
Legionella anisa NCTC 11974* | WDCM 00106 | R4601315 |
Enterococcus faecalis ATCC® 19433* | WDCM 00009 | R4601990 |
Enterococcus faecalis ATCC® 29212* | WDCM 00087 | R4607030 |
Pseudomonas aeruginosa ATCC® 27853* | WDCM 00025 | R4607060 |
Pseudomonas paraeruginosa ATCC® 9027* | WDCM 00026 | R4605210 |
Escherichia coli ATCC® 8739* | WDCM 00012 | R4607085 |
Escherichia coli ATCC® 25922* | WDCM 00013 | R4607050 |
Prepared Media for the Quality Control testing according to ISO 11133:2014 | ||
Tryptone Soya Agar (Caseine Soya Bean Digest Agar) | PO5073A |
* The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Remel Inc. is licensed to use these trademarks and sell products derived from ATCC® cultures. The identity, purity, and authenticity of the Licensed Products are exclusively the responsibility of Remel Inc., and not ATCC.
For Research Use Only. Not for use in diagnostic procedures.