Search
Deliver consistent results with a furnace you can depend on to attain your daily goals. Thermo Scientific Box and Tube Furnaces come in a wide range of offerings to accommodate ordinary and technical tasks alike, such as: temperature ranges up to 1200°C, temperature control options to meet your application needs, and embedded or open heating elements designed to keep samples safe while maintaining reliable temperature uniformity.
View our quick application guide for choosing a furnace that is suitable for the workload you need. Choose from a variety of temperature control options, heating elements and furnace formats to meet your analytical requirements.
Typically used for processing larger samples or to easily place and access samples. We offer a versatile selection of small, medium, and large box furnaces suitable for a variety of industrial and laboratory applications. Advanced engineering and specialized construction techniques include variable density insulation, double shell cabinets, long-life heating elements and side-swing doors (vertical or horizontal) or swing down doors.
| Thermolyne Benchtop Muffle Furnaces | ||||||
![]() |
![]() |
![]() |
![]() |
![]() |
![]() |
|
Heat range |
100 - 1200°C |
100 - 1100°C | 100 - 1093°C |
100 - 1200°C | 100 - 1200°C |
100 - 1100°C |
Available capacity |
16.4 - 55.3 L |
1.99 - 42.5 L | 45 L |
14 L |
2 - 5.8 L |
1.3 to 2.1 L |
Heating elements |
Embedded |
Embedded | Embedded |
Embedded |
Open |
Embedded |
Tube furnaces are typically used for processing small samples or heating in an inert atmosphere. The single and three-zoned construction is designed to create precise temperature control. Three-zone control enables the user to select a different temperature in each zone (e.g. for gas applications or material experiments). Some models offer split-hinge design, which easily allows you to change the tube.
![]() |
![]() |
|
|
Heat range |
100 - 1200°C |
100 - 1100°C |
100 - 1100°C |
Available capacity |
30.5, 61, or 91.4 cm / 12, 24, or 36 in. |
61 or 91.4 cm / 24 or 36 in. |
30.5 cm / 12 in. |
Heating elements |
Open |
Embedded |
Open |
|
Description |
Dimensions |
Cat. no. |
|---|---|---|---|
Thermolyne premium large muffle furnaces |
Critical to operation - an appropriate exhaust must be installed with your furnace to remove smoke and gases produced during an ashing procedure. Simplify installation when you order this kit. |
Kit includes tubing, brackets, and screws. |
AY408X1A |
Thermolyne largest tabletop muffle furnaces |
|||
Lindberg/Blue M 1100 °C tube furnaces* –STF55346 and STF55666 series models
|
Increase the flexibility of your applications with additional sized tube adapters. Tube adapters hold process tubes of a certain size in place to ensure the tubes don't contact the sides of the tube furnace, protecting the furnace heating elements. |
3-in. adapter |
59545 |
3-in. adapter |
59555 |
||
4-in. adapter |
59556 |
||
Blank (solid) adapter |
59549 |
||
Lindberg/Blue M 1200 °C split-hinge tube furnaces* – HTF55122, HTF55322, HTF55342, and HTF55347 series models |
0.75-in adapter |
59510 |
|
1-in. adapter |
59511 |
||
Blank (solid) adapter |
59519 |
||
1-in. adapter |
59521 |
||
1.5-in. adapter |
59522 |
||
2-in. adapter |
59523 |
||
2.5-in. adapter |
59524 |
||
3-in. adapter |
59525 |
||
Blank (solid) adapter |
59529 |
||
*Additional adapters may be available. Contact your local representative or customer support to order the following: adapters for Lindberg/Blue M 1100 °C furnace model STF55346: 1-in. (Cat. No. 59541TA); adapters for Lindberg/Blue M 1100 °C furnace model STF55666: 6-in. (Cat. No. 59558TA), blank (solid) adapter (Cat. No. 59559TA); and adapters for Lindberg/Blue M 1200 °C split-hinge furnace model HTF55667: 3-in. (Cat. No. 59535TA), 4-in. (Cat. No. 59536TA), 5-in. (Cat. No. 59537TA), 6-in. (Cat. No. 59538TA), blank (solid) adapter (Cat. No. 59539TA).
Replacement part |
For use with |
Description |
Dimensions |
Cat. no. |
F47900 Series |
Hearth trays and hearth plates sit in the bottom of the furnace and protect the bottom heating element during operation. |
15.2 x 14.3 x 0.95 cm./ 5.98 x 5.63 x 0.37 in. |
PH479X1 |
|
F80000 Series
|
25.4 x 19.3 x 0.95 cm. / 10 x 7.6 x 0.38 in.
|
PH480X1 |
||
Ceramic shelves expand interior usable sample space without interfering with max operation temperature. |
17.4 x 17.3 x 1.2 cm. / 6.85 x 6.81 x 0.47 in.
|
SH480X1 |
Replacement part |
Description |
Dimensions |
Cat. no |
Order extra trays designed to custom configure your furnace. Trays hold crucibles of various sizes, meant to sit on shelves or hearth trays when in use. |
32.4 x 25.4 x 17.1 cm. / 12.75 x 10 x 6.75 in.
|
TY408X2A |
|
Hearth trays and hearth plates sit in the bottom of the furnace and protect the bottom heating element during operation. |
8.2 x 10.1 x 1.27 cm. / 3.3 x 4 x 0.5 in. |
PHX1 |
|
25.4 x 19.3 x 0.95 cm. / 10 x 7.6 x 0.38 in. |
PH177X1 |
||
Stainless steel shelves expand interior usable sample space, but limit the max operation temperature to 975°C. If higher heat is required, use ceramic shelves. |
32.5 x 21.1 cm. / 12.8 x 8.3 in.
|
SH408X1 |
|
For use with crucible trays, to pull a hot tray in and out of the furnace. |
|
HN408X2A |
|
Holds shelves in place and allows flexibility of shelf placement. |
|
JSX16 |
Replacement part |
Description |
Dimensions |
Cat. no |
Order extra trays designed to custom configure your furnace. Trays hold crucibles of various sizes, meant to sit on shelves or hearth trays when in use. |
32.4 x 25.4 x 17.1 cm. / 12.75 x 10 x 6.75 in.
|
TY408X2A |
|
Hearth trays and hearth plates sit in the bottom of the furnace and protect the bottom heating element during operation. |
17.1 x 14.9 x 1.9 cm. / 6.75 x 5.9 x 0.75 in. |
PH146X1 |
|
Refractory shelves expand interior usable sample space without interfering with max operation temperature. |
35.2 x 25.4 x 1.27 cm. / 13.87 x 10 x 0.56 in. |
SH412X1 |
|
For use with crucible trays, to pull a hot tray in and out of the furnace. |
|
HN408X2A |
*Additional replacement parts may be available. Contact your local representative or customer support to order the following: hearth tray for Thermolyne small benchtop muffle furnace model FB1300 (Cat. No. PH44X1); hearth tray for Thermolyne small benchtop muffle furnace model FB1400 (Cat. No. PH48X1); hearth tray for Thermolyne industrial benchtop muffle furnace (Cat. No. PHX2).
Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services