Introduction
Dynabeads® M-280 Sheep anti-Mouse IgG are uniform, superparamagnetic, polystyrene beads with affinity purified sheep anti-mouse IgG covalently bound to the bead surface. These secondary polyclonal antibodies bind both heavy and light chain to mouse IgG 1, IgG 2a and IgG 2b. They may have low reactivity towards mouse IgG 3 and IgM. Human cross reactivity is minimal. The beads are supplied as a suspension containing 6-7 x 10 8 beads/ml approx. (10 mg/ml) in phosphate buffered saline (PBS) pH 7.4 with 0.1% bovine serum albumin (BSA) and 0.02% sodium azide (NaN 3).
Principle
Dynabeads® M-280 Sheep anti-Mouse IgG is designed as a solid support for simple and efficient binding of immunoglobulins (Ig) or other target molecules. The sheep anti-mouse IgG bound to the surface of the beads will bind defined antigens via a mouse primary antibody.
The size of the beads (2.8 μm) makes them particularly suitable for isolation of antibodies (Ab) and their target proteins. The beads can also be used for cell isolation, but the larger 4.5 μm Dynabeads® are more frequently used for this application.
Using a magnet (Dynal® MPC™, Magnetic Particle Concentrator), the beads allow isolation and subsequent handling of target molecules in a highly specific manner. The beads are added directly to the sample containing your target antibody/antigen. After a short incubation allowing affinity capture of the target, the beads are pulled to the side of the test-tube by the use of a magnet, allowing aspiration of unbound material. Alternatively, an indirect approach can be of benefit when the concentration of antibody is low, the antibody-antigen affinity is weak or the binding kinetics is slow. In the indirect approach, the primary antibody is allowed to bind to the target in suspension prior to addition of the Dynabeads® M-280 Sheep anti-Mouse IgG.
Magnetic separation facilitates easy washing and concentration of the isolated target. The target molecule can be eluted off the beads with conventional elution methods, or used in further downstream applications while still attached to the beads. If your downstream application involves purification and elution of a target protein, you might want to crosslink your primary antibody to the sheep anti-mouse IgG on the Dynabeads® before immunoprecipitation to prevent co-elution of the primary antibody.
TOP
Materials
Additional Material Needed
  • Magnetic device (Dynal® MPC, Magnetic Particle Concentrator).
  • Mixing/rotation device
  • Vortex mixer
  • Test tubes and pipettes
  • Buffers/solutions
  • Primary antibody
Recommended Buffers/Solutions
All reagents used should be analytical grade. Isotonic (PBS) pH 7.2-7.6 Invitrogen Dynal® recommends the following PBS (pH 7.4):
NaH 2PO 4 x H 2O 0.16 g
Na 2HPO 4 x 2 H 2O 0.98 g
NaCl 8.10 g
Distilled water to 1 litre
PBS/BSA: Add 0.1% BSA fraction V (final conc.) to PBS. 0.02% NaN 3 may be added as a preservative for storage, if needed.
Protocols
Instructions for use
Washing Procedure
The Dynabeads® M-280 Sheep anti-Mouse IgG should be washed before use to remove preservatives. The washing procedure is facilitated by the use of a magnet. Make sure the beads are thoroughly resuspended to a homogeneous solution before use (in step 2 below).
  1. Resuspend the Dynabeads® M-280 Sheep anti-Mouse IgG thoroughly in the vial by vortexing and/or shaking.
  2. Transfer the required amount of resuspended beads into a tube.
  3. Place the tube on the magnet for 2 minutes and pipette off the supernatant. Avoid touching the inside wall of the tube (where the beads attract to the magnet) with the pipette tip.
  4. Remove the tube from the magnet, and resuspend the pellet in an excess volume of washing buffer.
  5. Repeat step 3 and again resuspend the washed beads in washing buffer.
The washed Dynabeads® M-280 Sheep anti-Mouse IgG can be resuspended in any chosen volume. A small volume should be chosen (e.g. the volume originally pipetted from the vial) so that a final bead concentration of 1-2 x 107 beads/ml can be achieved for efficient target isolation.
Separation of Target Ig
  1. Resuspend the washed Dynabeads® M-280 Sheep anti-Mouse IgG well by vortexing or shaking
  2. Add the sample containing your target-Ig to the washed beads. Approx. 0.1-1 μg Ig/107 beads may be sufficient for binding.
  3. Incubate with slow tilt rotation mixing for 30 min or up to 24 hours at 2-8°C.
  4. Place the test tube on the magnet for 2 minutes and pipette off the supernatant.
  5. Remove the test tube from the magnet, add washing buffer and resuspend.
  6. Repeat steps 4 and 5 twice, place the tube on the magnet and remove the supernatant. The bound and purified Ig is now ready to be eluted off the beads, or the Ig-coated beads can be used for immunoprecipitation. The latter is done by either adding the coated beads directly to a new sample containing the target protein, or by first covalently crosslinking the primary Ig to the sheep anti-mouse IgG on the beads.
Elution of Isolated Ig
Eluting the isolated Ig off the Dynabeads® M-280 Sheep anti-Mouse IgG is – in this example – performed by lowering pH using 0.1 M citrate (pH 2-3) as the elution buffer. The degree of acidity needed will depend on the specific Ig, but at pH 3.1 most Ig will be eluted off.
  1. Add 0.1 M citrate (pH 2-3) to Dynabeads® M-280 Sheep anti-Mouse IgG with immobilized Ig.
  2. Mix well by tilting and rotation for 2 minutes.
  3. Place the test tube on a magnet and transfer the supernatant, containing purified Ig, to a clean tube.
  4. Again add 0.1 M citrate (pH 2-3) to the beads to elute any remaining Ig.
  5. Mix well by tilting and rotation for 2 minutes.
  6. Place the test tube on a magnet, pipette off the eluate and pool the supernatants containing, pure Ig.
Dynabeads® M-280 Sheep anti-Mouse IgG where the Ig has been eluted off may be reused at least five times. For re-use after elution, the beads should immediately be brought to neutral pH using a Na-phosphate buffer. For storage, the beads should be resuspended in a PBS/BSA buffer.
Immunoprecipitation
Bound Ig will be co-eluted along with the target using different elution methods (see section below). When isolating antigens, for SDS-PAGE followed by Western blotting or autoradiography, the presence of Ig may not disturb the detection system. For other applications such as protein purification or amino acid sequencing, co-elution of the Ig is not desired. If the presence of Ig will not disturb your detection system, go directly to section below. For applications where co-elution of the Ig is not desired, your primary Ig can be crosslinked to the sheep anti-mouse IgG as described below. If trace amounts of Ig is not crosslinked, this can be removed prior to immunoprecipitation by following the procedure described in above. After isolation and mild elution of the target protein, the Ig-coated beads may be reused for immunoprecipitation. If the Ig-coated beads are to be reused, crosslinking is necessary.
Crosslinking Ig to the Beads
The protocol presented here uses 0.2 M triethanolamine pH 8.2. Other non-amine containing buffers with pH 7-9 can also be used.
  1. Add 1 ml 0.2 M triethanolamine, pH 8.2 to the Dynabeads® M-280 Sheep anti-Mouse IgG with immobilized Ig. Wash twice using a magnet and 0.2 M triethanolamine, pH 8.2 as the washing buffer (see section above).
  2. Resuspend the beads in 1 ml of 20 mM DMP (dimethyl pimelimidate dihydrochloride, Pierce #21666) in 0.2 M triethanolamine, pH 8.2 (5.4 mg DMP/ml buffer). This crosslinking solution must be prepared immediately before adding to the beads.
  3. Incubate with rotational mixing for 30 minutes at 20°C. Place the tube on the magnet and discard the supernatant.
  4. Remove the tube from the magnet and stop the reaction by resuspending the beads in 1 ml of 50 mM Tris, pH 7.5 and incubate for 15 minutes with rotational mixing.
  5. Place the tube on the magnet and discard the supernatant.
  6. Wash the now crosslinked Dynabeads® 3 times with 1 ml PBS/BSA by the use of a magnet. Pre-elution is optional. Resuspend the beads in chosen volume or add directly to antigen-containing solution. Invitrogen Dynal® can not guarantee the full recovery of your Ig activity, as this varies from Ig to Ig.
Antigen-Binding to Ig-Coated Beads
Binding of protein or other antigen to primary Ig-coated Dynabeads® is dependent on the concentration of the beads, antigen concentration, the affinity of the immobilized Ig and time. Binding is performed at 2-8°C from 10 minutes to 1 hour. Equilibrium antibody-antigen is reached after approximately 1 hour.
  1. Add sample containing antigen to the beads. For a 100 kD protein, use a volume containing approximate 25 μg target antigen/ml beads to assure an excess of antigen. If dilution of antigen is necessary, PBS or 0.1 M phosphate buffer (pH 7-8) can be used as dilution buffer.
  2. Incubate with tilting and rotation for one hour. (Incubation times as low as 10 minutes can be used with concentrated protein samples in volumes close to what was originally pipetted from the product vial).
  3. Place the tube on the magnet for 2 minutes to collect the Ig-coated Dynabeads®-target complex at the tube wall. For viscous samples, double the time on the magnet. Pipette off the supernatant.
  4. Wash the beads 3 times using 1 ml PBS each time and change buffers by the use of a magnet (see section above).
Target Protein Elution
Conventional elution methods can be applied for the elution of target antigen from the Dynabeads®, and one of the major advantages using Dynabeads® in protein/Ig isolation is the possibility to elute in small volumes. Low pH (2.8–3.5), change in ionic strength, affinity elution, electrophoresis, polarity reducing agents, deforming eluents can be applied, or even boiling the bead-target complex in SDS-PAGE application buffer for direct characterization of protein on SDS-PAGE. The method of choice depends on the Ig’s affinity for the specific target protein, stability of target protein and downstream applications and detection methods. Most proteins will be eluted at pH 3.1 following the procedure described under Dynabeads® Ig elution procedure (see section above). Some protein functionality might be lost under these conditions. If maintaining functionality of the target protein is important, try milder elution conditions first such as high salt (e.g. 2M NaCI) or step-wise elution reducing pH from 6 down to 3. This is also recommended if the bead-bound ligand must remain functional to allow reuse of the Dynabeads®.
TOP
General Information
Invitrogen Dynal® AS complies with the Quality System Standards ISO 9001:2000 and ISO 13485:2003.
Product Characteristics
Typical bead-characteristics for any given lot of this product: Diameter: 2.8 μm Specific surface area: 4-8 m2/g Density: 1.4 g/cm3 Certificate of Analysis (CoA) is available upon request. Material Safety Data Sheet (MSDS) ism available at www.invitrogen.com
TOP
References
  1. Harcourt G. et al. Detection of a-subunit isoforms in human muscle acetylcholine receptor by specific T cell from a myasthenia gravis patient. Proc.R.Soc.Lond.B 1993;254:1-6
  2. Löliger C, Rühlmann E and Kühnl P. A rapid and sensitive immunoassay for antibodies against alloantigens on human platelet glycoproteins (BIPA). J.Immunol.Meth.1993;158:197-200
  3. Ostermann J. et al. Stepwise assembly of functionally active transport vesicles. Cell 1993;75: 1015-1025
  4. Astier A. et al. Human epidermal Langerhans cells secrete a soluble receptor for IgG (Fc gamma RII/CD32) that inhibits the binding of immune complexes to Fc gamma R+ cells. J.Immunol. 1994;152:201-212
  5. Grinde B, Jonassen T and Ushijima H. Sensitive detection of group A rotaviruses by immunomagnetic separation and RT-PCR. J.Virol. Methods 1995;55:327-338
  6. Rashkovetsky LG. et al. Automated microanalysis using magnetic beads with commercial capillary electrophoretic instrumentation. J.Chromatography 1997;A781:197-204
  7. Calhoun BC and Goldenring JR. Two Rab proteins, vesicle-associated membrane protein 2 (VAMP-2) and secretory carrier membrane proteins (SCAMPs), are present on immunoisolated parietal cell tubulovesicles. Biochem J. 1997;Jul 15;325 (Pt 2):559-64
  8. Woods AJ. et al. Paxillin associates with poly(A)-binding protein 1 at the dense endoplasmic reticulum and the leading edge of migrating cells. J Biol Chem. 2002;Feb 22;277(8):6428-37. Epub 2001 Nov 09
  9. Hofmann I. et al. Symplekin, a constitutive protein of karyo- and cytoplasmic particles involved in mRNA biogenesis in Xenopus laevis oocytes. Mol Biol Cell. 2002;May;13(5):1665-76
  10. Hamm J. et al. Bi-functional, substrate mimicking RNA inhibits MSK-1 mediated cAMP response element-binding protein phosphorylation and reveals magnesium ion-dependent conformational changes of the kinase. J.Biol.Chem. 2002;277(48):45793-45802
  11. Oh P. et al., Dynamin at the neck of caveolae mediates their budding to form transport vesicles by GTP-driven fission from the plasma membrane of endothelium. J.Cell Biol. 1998;141(1):101-114
TOP
112.01D_02D.indd    Rev 006    5-May-2007