Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Inorganic Salts | |||
| Sodium Chloride (NaCl) | 58.0 | 5000.0 | 86.206894 |
| Other Components | |||
| SELECT Peptone 140 (pancreatic digest of casein) | 10000.0 | Infinity | |
| SELECT Yeast Extract autolyzed low sodium | 5000.0 | Infinity | |
LB Agar, powder (Lennox L Agar)
Cat No:
22700025
,
22700041
LB Broth (1X) liquid
Cat No:
10855021