Search
INTENDED USE: A buffered, enriched medium to obtain high yields of plasmid DNA from transformed E. coli (1). INSTRUCTIONS FOR USE: Suspend 47 g in 1 L of demineralized water. Add 4 ml of glycerol. Autoclave for 15 min at 121°C. RECOMMENDED STORAGE CONDITION: 15°C TO 30°C.
Catalog Number(s)
22711022| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Inorganic Salts | |||
| Dipotassium hydrogen phosphate | 174.176 | 9.4 | 0.0539684 |
| Potassium dihydrogen phosphate | 136.086 | 2.2 | 0.016166247 |
| Other Components | |||
| SELECT Peptone 140 (pancreatic digest of casein) | 11.8 | Infinity | |
| SELECT Yeast Extract autolyzed low sodium | 23.6 | Infinity | |
1. Tartof, K.D. and Hobbs, C.A. (1987) Focus® 9:2, 12.
2-YT Broth
Cat No:
22712020
Gibco® BG-11 Media: Optimized for Cyanobacteria
Cat No:
a1379901
,
A1379902
TAP Growth Media, optimized for Chlamydomonas culture
Cat No:
A1379801
,
A1379802