Search
Catalog Number(s)
A1048801| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Amino Acids | |||
| L-Arginine | 174.0 | 127.0 | 0.72988504 |
| L-Asparagine | 132.0 | 60.0 | 0.45454547 |
| L-Cystine | 240.0 | 24.0 | 0.1 |
| L-Glutamine | 146.0 | 292.0 | 2.0 |
| L-Histidine hydrochloride-H2O | 210.0 | 42.0 | 0.2 |
| L-Isoleucine | 131.0 | 52.0 | 0.39694658 |
| L-Leucine | 131.0 | 131.0 | 1.0 |
| L-Lysine hydrochloride | 183.0 | 73.0 | 0.3989071 |
| L-Methionine | 149.0 | 15.0 | 0.10067114 |
| L-Phenylalanine | 165.0 | 32.0 | 0.19393939 |
| L-Serine | 105.0 | 42.0 | 0.4 |
| L-Threonine | 119.0 | 48.0 | 0.40336135 |
| L-Tryptophan | 204.0 | 10.0 | 0.04901961 |
| L-Tyrosine | 181.0 | 36.0 | 0.19889502 |
| L-Valine | 117.0 | 46.0 | 0.3931624 |
| Vitamins | |||
| Biotin | 244.0 | 0.1 | 4.0983607E-4 |
| Choline chloride | 140.0 | 56.0 | 0.4 |
| D-Calcium pantothenate | 477.0 | 1.0 | 0.002096436 |
| Folic Acid | 441.0 | 2.2 | 0.0049886624 |
| Niacinamide | 122.0 | 1.0 | 0.008196721 |
| Pyridoxal hydrochloride | 204.0 | 1.0 | 0.004901961 |
| Riboflavin | 376.0 | 0.1 | 2.6595744E-4 |
| Thiamine hydrochloride | 337.0 | 1.0 | 0.002967359 |
| Vitamin B12 | 1355.0 | 1.36 | 0.0010036901 |
| i-Inositol | 180.0 | 36.0 | 0.2 |
| Inorganic Salts | |||
| Calcium Chloride (CaCl2-2H2O) | 147.0 | 265.0 | 1.8027211 |
| Ferric Chloride 6H2O | 270.0 | 0.54 | 0.002 |
| Magnesium Chloride (MgCl2-6H2O) | 203.0 | 183.0 | 0.9014778 |
| Magnesium Sulfate (MgSO4-7H2O) | 246.0 | 25.0 | 0.101626016 |
| Potassium Chloride (KCl) | 75.0 | 400.0 | 5.3333335 |
| Sodium Bicarbonate (NaHCO3) | 84.0 | 2200.0 | 26.190475 |
| Sodium Chloride (NaCl) | 58.0 | 6800.0 | 117.24138 |
| Sodium Phosphate monobasic (NaH2PO4-H2O) | 138.0 | 150.0 | 1.0869565 |
| Zinc sulfate (ZnSO4-7H2O) | 288.0 | 0.14 | 4.8611112E-4 |
| Other Components | |||
| D-Glucose (Dextrose) | 180.0 | 2000.0 | 11.111111 |
| Linoleic Acid | 280.0 | 0.084 | 2.9999999E-4 |
| Lipoic Acid | 206.0 | 0.2 | 9.708738E-4 |
| Putrescine 2HCl | 161.0 | 0.16 | 9.937888E-4 |
| Sodium Pyruvate | 110.0 | 110.0 | 1.0 |
10373 - MEM, Richter's, no phenol red
Cat No:
10373017
A10489 - MEM, Richter’s, no Gentamycin
Cat No:
a1048901