Search
Without Calcium.
A sterile, liquid medium for the culture of human epidermal melanocytes. Medium 254CF is Medium 254 prepared without calcium chloride. Calcium chloride is provided as a separate component with each bottle of medium. This basal medium requires the addition of HMGS (S-002-5) or HMGS-2 (S-016-5) prior to use. Below is the composition list. The concentrations are proprietary.
Catalog Number(s)
M254CF500| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Amino Acids | |||
| Glycine | 75.0 | confidential | n/a |
| L-Alanine | 89.0 | confidential | n/a |
| L-Alanyl-L-Glutamine | 217.0 | confidential | n/a |
| L-Arginine hydrochloride | 211.0 | confidential | n/a |
| L-Asparagine-H2O | 150.0 | confidential | n/a |
| L-Aspartic acid | 133.0 | confidential | n/a |
| L-Cysteine | 121.0 | confidential | n/a |
| L-Glutamic Acid | 147.0 | confidential | n/a |
| L-Histidine hydrochloride-H2O | 210.0 | confidential | n/a |
| L-Isoleucine | 131.0 | confidential | n/a |
| L-Leucine | 131.0 | confidential | n/a |
| L-Lysine hydrochloride | 183.0 | confidential | n/a |
| L-Methionine | 149.0 | confidential | n/a |
| L-Phenylalanine | 165.0 | confidential | n/a |
| L-Proline | 115.0 | confidential | n/a |
| L-Serine | 105.0 | confidential | n/a |
| L-Threonine | 119.0 | confidential | n/a |
| L-Tryptophan | 204.0 | confidential | n/a |
| L-Tyrosine | 181.0 | confidential | n/a |
| L-Valine | 117.0 | confidential | n/a |
| Vitamins | |||
| Choline chloride | 140.0 | confidential | n/a |
| D-Pantothenic Acid | 238.0 | confidential | n/a |
| Folic Acid | 441.0 | confidential | n/a |
| Myo-Inositol | 180.0 | confidential | n/a |
| Niacinamide | 122.0 | confidential | n/a |
| Pyridoxal hydrochloride | 206.0 | confidential | n/a |
| Riboflavin | 376.0 | confidential | n/a |
| Thiamine hydrochloride | 337.0 | confidential | n/a |
| Vitamin B12 | 1355.0 | confidential | n/a |
| d-Biotin | 244.0 | confidential | n/a |
| Inorganic Salts | |||
| Ammonium Metavanadate NaVO3 | 117.0 | confidential | n/a |
| Ammonium Molybdate ((NH4)6Mo7O24-4H2O) | 1236.0 | confidential | n/a |
| Cupric sulfate (CuSO4-5H2O) | 250.0 | confidential | n/a |
| Ferric sulfate (FeSO4-7H2O) | 278.0 | confidential | n/a |
| Magnesium Chloride (MgCl2-6H2O) | 203.0 | confidential | n/a |
| Manganese Sulfate (MnSO4-H20) | 169.0 | confidential | n/a |
| Nickelous Chloride NiCl2 6H2O | 238.0 | confidential | n/a |
| Potassium Chloride (KCl) | 75.0 | confidential | n/a |
| Sodium Acetate (NaCH3COO.3H2O) | 136.0 | confidential | n/a |
| Sodium Bicarbonate (NaHCO3) | 84.0 | confidential | n/a |
| Sodium Chloride (NaCl) | 58.0 | confidential | n/a |
| Sodium Meta Silicate Na2SiO3 9H2O | 284.0 | confidential | n/a |
| Sodium Phosphate dibasic (Na2HPO4-7H2O) | 268.0 | confidential | n/a |
| Sodium Selenite (Na2SeO3) | 173.0 | confidential | n/a |
| Tin Chloride (SnCl2.2H2O) | 226.0 | confidential | n/a |
| Zinc sulfate (ZnSO4-7H2O) | 288.0 | confidential | n/a |
| Other Components | |||
| Adenine.HCl | 172.0 | confidential | n/a |
| D-Glucose (Dextrose) | 180.0 | confidential | n/a |
| DL-alpha-Lipoic Acid | 206.0 | confidential | n/a |
| Ethanolamine | 61.0 | confidential | n/a |
| HEPES | 238.0 | confidential | n/a |
| O-Phosphorylethanolamine | 141.0 | confidential | n/a |
| Phenol Red | 376.4 | confidential | n/a |
| Putrescine 2HCl | 161.0 | confidential | n/a |
| Sodium Pyruvate | 110.0 | confidential | n/a |
| Thymidine | 242.0 | confidential | n/a |
Medium 106
Cat No:
M106500
Medium 131
Cat No:
M131500
Medium 154
Cat No:
M154500
Medium 154
Cat No:
M154CF500
Medium 154
Cat No:
M154CFPRF500
EpiLife® Medium
Cat No:
MEPI500CA
EpiLife® Medium
Cat No:
MEPICF500
EpiLife® Medium
Cat No:
MEPICFPRF500
Medium 231
Cat No:
M231500
Medium 200
Cat No:
m200500
Medium 200
Cat No:
M200PRF500
Medium 254
Cat No:
M254500
Medium 171
Cat No:
M171500