Amino Acids |
Glycine |
75.0 |
13.6 |
0.18133333 |
Hydroxy L-proline |
131.0 |
4.09 |
0.031221375 |
L-Alanine |
89.0 |
31.6 |
0.3550562 |
L-Arginine hydrochloride |
211.0 |
31.16 |
0.14767772 |
L-Asparagine-H2O |
150.0 |
9.17 |
0.061133333 |
L-Aspartic acid |
133.0 |
9.91 |
0.074511275 |
L-Cystine 2HCl |
313.0 |
14.0 |
0.044728436 |
L-Glutamic Acid |
147.0 |
8.26 |
0.05619048 |
L-Glutamine |
146.0 |
135.7 |
0.92945206 |
L-Histidine hydrochloride-H2O |
210.0 |
26.65 |
0.12690476 |
L-Isoleucine |
131.0 |
18.04 |
0.13770993 |
L-Leucine |
131.0 |
20.44 |
0.15603054 |
L-Lysine hydrochloride |
183.0 |
38.43 |
0.21000001 |
L-Methionine |
149.0 |
4.44 |
0.029798659 |
L-Ornithine HCl |
169.0 |
9.41 |
0.055680472 |
L-Phenylalanine |
165.0 |
16.53 |
0.100181825 |
L-Proline |
115.0 |
6.13 |
0.05330435 |
L-Serine |
105.0 |
10.75 |
0.102380954 |
L-Taurine |
125.0 |
4.18 |
0.033439998 |
L-Threonine |
119.0 |
18.93 |
0.15907563 |
L-Tryptophan |
204.0 |
17.5 |
0.085784316 |
L-Tyrosine disodium salt dihydrate |
181.0 |
21.0 |
0.1160221 |
L-Valine |
117.0 |
25.0 |
0.21367522 |
Vitamins |
Biotin |
244.0 |
0.025 |
1.0245902E-4 |
Choline chloride |
140.0 |
1.25 |
0.008928572 |
D-Calcium pantothenate |
477.0 |
0.025 |
5.2410902E-5 |
Folic Acid |
441.0 |
0.025 |
5.6689343E-5 |
Menadione (Vitamin K3) |
172.0 |
0.025 |
1.4534884E-4 |
Niacinamide |
122.0 |
0.0625 |
5.1229505E-4 |
Nicotinic acid (Niacin) |
123.0 |
0.0625 |
5.0813006E-4 |
Para-Aminobenzoic Acid |
137.0 |
0.125 |
9.1240875E-4 |
Pyridoxal hydrochloride |
204.0 |
0.0625 |
3.0637256E-4 |
Pyridoxine hydrochloride |
206.0 |
0.0625 |
3.0339806E-4 |
Riboflavin |
376.0 |
0.025 |
6.648936E-5 |
Thiamine hydrochloride |
337.0 |
0.025 |
7.4183976E-5 |
Vitamin A (acetate) |
286.0 |
0.25 |
8.7412586E-4 |
Vitamin B12 |
1355.0 |
10.0 |
0.007380074 |
Vitamin D2 (Calciferol) |
397.0 |
0.25 |
6.297229E-4 |
alpha Tocopherol phos. Na salt |
702.0 |
0.025 |
3.5612535E-5 |
i-Inositol |
180.0 |
0.125 |
6.9444446E-4 |
Inorganic Salts |
Calcium Chloride (CaCl2) (anhyd.) |
111.0 |
200.0 |
1.8018018 |
Magnesium Sulfate (MgSO4) (anhyd.) |
120.0 |
97.67 |
0.8139166 |
Potassium Chloride (KCl) |
75.0 |
400.0 |
5.3333335 |
Sodium Bicarbonate (NaHCO3) |
84.0 |
2200.0 |
26.190475 |
Sodium Chloride (NaCl) |
58.0 |
6800.0 |
117.24138 |
Sodium Phosphate monobasic (NaH2PO4-H2O) |
138.0 |
140.0 |
1.0144928 |
Ribonucleosides |
5-Methylcytosine |
225.0 |
0.1 |
4.4444445E-4 |
Deoxyribonucleosides |
2'Deoxyadenosine |
251.0 |
10.0 |
0.03984064 |
2'Deoxycytidine HCl |
264.0 |
10.0 |
0.03787879 |
2'Deoxyguanosine |
267.0 |
10.0 |
0.037453182 |
Thymidine |
242.0 |
10.0 |
0.041322313 |
Coenzymes |
CoA (Coenzyme A) |
768.0 |
2.5 |
0.0032552083 |
DPN (diphosphopyridine nucleotide) |
663.0 |
7.0 |
0.01055807 |
FAD (flavin adenine dinucleotide) |
786.0 |
1.0 |
0.0012722646 |
TPN (triphosphopyridine nucleotide-Na) |
743.0 |
1.0 |
0.001345895 |
TPP Co-carboxylase |
461.0 |
1.0 |
0.0021691974 |
UTP (uridine triphosphate 3Na-2H20) |
586.0 |
1.0 |
0.0017064846 |
Reducing Agents |
Ascorbic Acid |
176.0 |
50.0 |
0.2840909 |
Glutathione, monosodium |
329.0 |
10.0 |
0.030395137 |
Other Components |
D-Glucosamine HCl |
216.0 |
3.85 |
0.017824074 |
D-Glucose (Dextrose) |
180.0 |
1000.0 |
5.5555553 |
D-Glucuronolactone |
176.0 |
1.8 |
0.010227272 |
L-alpha-Amino-n-butyric acid |
103.0 |
5.5 |
0.053398058 |
Phenol Red |
376.4 |
20.0 |
0.053134963 |
Sodium Acetate |
136.0 |
50.0 |
0.36764705 |
Sodium glucuronate-H2O |
236.0 |
1.8 |
0.0076271184 |
Tween 80® |
|
12.5 |
Infinity |