Technical Resources - Media Formulations
15210 - KaryoMAX(R) Colcemid Solution in HBSS
  • Catalog Number(s)
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Calcium Chloride (CaCl2) (anhyd.)111.0100.00.9009009
Magnesium Chloride (MgCl2-6H2O)203.0100.00.49261084
Magnesium Sulfate (MgSO4-7H2O)246.0100.00.40650406
Potassium Chloride (KCl)75.0400.05.3333335
Potassium Phosphate monobasic (KH2PO4)142.060.00.4225352
Sodium Bicarbonate (NaHCO3)84.0350.04.1666665
Sodium Chloride (NaCl)58.08000.0137.93103
Sodium Phosphate dibasic (Na2HPO4) anhydrous142.090.00.63380283
Other Components
Colcemid(TM)371.010.00.026954178
D-Glucose (Dextrose)180.01000.05.5555553
Phenol Red398.0140.00.3517588
Reference:
Related Products:
15212 - KaryoMAX(R) Colcemid Solution in PBS
Cat No: 15212012