cAMP-Screen Direct System protocol
With the cAMP-Screen Direct™ Immunoassay System, cells are grown directly in the cAMP quantitation assay plate without subsequent lysate transfer. This eliminates the need for a separate culture plate, thereby reducing total assay costs, simplifying automation requirements for assay execution, and offering increased intra-assay accuracy.
With the cAMP-Screen Direct™ Immunoassay System, cells are grown directly in the cAMP quantitation assay plate without subsequent lysate transfer. This eliminates the need for a separate culture plate, thereby reducing total assay costs, simplifying automation requirements for assay execution, and offering increased intra-assay accuracy.
Related Products
Related Images