Rabbit IgG (H+L) Highly Cross-Adsorbed Antibody (A-31577) in IF
Immunofluorescence analysis of Goat anti-Rabbit IgG (H+L) Secondary Antibody, Alexa Fluor 635 was performed using HepG2 cells stained with alpha-1 antitrypsin Rabbit Polyclonal Primary Antibody (PA516661). The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton? X-100 for 10 minutes, blocked with 1% BSA for 1 hour and labeled with Rabbit primary antibody (1:250 dilution) 3 hours at room temperature. Goat anti-Rabbit IgG (H+L) Secondary Antibody, Alexa Fluor 635 (A31577) was used at a concentration of 4ug/ml in phosphate buffered saline containing 0.2 % BSA for 45 minutes at room temperature, for detection of alpha-1 antitrypsin in the cytoplasm (Panel a: red). Nuclei (Panel b: blue) were stained with DAPI in SlowFade® Gold Antifade Mountant (S36938). F-actin was stained with Alexa Fluor® 488 Phalloidin (A12379, 1:300) (Panel c: green). Panel d represents the composite image. No nonspecific staining was observed with the secondary antibody alone (panel f), or with an isotype control (panel e). The images were captured at 60X magnification.
Immunofluorescence analysis of Goat anti-Rabbit IgG (H+L) Secondary Antibody, Alexa Fluor 635 was performed using HepG2 cells stained with alpha-1 antitrypsin Rabbit Polyclonal Primary Antibody (PA516661). The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton? X-100 for 10 minutes, blocked with 1% BSA for 1 hour and labeled with Rabbit primary antibody (1:250 dilution) 3 hours at room temperature. Goat anti-Rabbit IgG (H+L) Secondary Antibody, Alexa Fluor 635 (A31577) was used at a concentration of 4ug/ml in phosphate buffered saline containing 0.2 % BSA for 45 minutes at room temperature, for detection of alpha-1 antitrypsin in the cytoplasm (Panel a: red). Nuclei (Panel b: blue) were stained with DAPI in SlowFade® Gold Antifade Mountant (S36938). F-actin was stained with Alexa Fluor® 488 Phalloidin (A12379, 1:300) (Panel c: green). Panel d represents the composite image. No nonspecific staining was observed with the secondary antibody alone (panel f), or with an isotype control (panel e). The images were captured at 60X magnification.
Related Products
Related Images