Ki-67 Antibody (47-5698-82) in Flow
Intracellular staining of mouse splenocytes unstimulated (left) or stimulated for 2 days with immobilized Anti-Mouse CD3 Functional Grade (Product ## 16-0031) (right) with Anti-Human/Mouse CD45R (B220) FITC (Product ## 11-0452) and 0.03 µg of Anti-Mouse/Rat Ki-67 APC-eFluor® 780 using the Foxp3/Transcription Factor Buffer Set (Product ## 00-5523) and protocol. Cells in the lymphocyte gate were used for analysis.
Intracellular staining of mouse splenocytes unstimulated (left) or stimulated for 2 days with immobilized Anti-Mouse CD3 Functional Grade (Product ## 16-0031) (right) with Anti-Human/Mouse CD45R (B220) FITC (Product ## 11-0452) and 0.03 µg of Anti-Mouse/Rat Ki-67 APC-eFluor® 780 using the Foxp3/Transcription Factor Buffer Set (Product ## 00-5523) and protocol. Cells in the lymphocyte gate were used for analysis.
Related Products
Related Images