Search
Rechercher
Trinitrobenzene sulfonate (TNBS) reacts with primary amines molecules to from an orange-colored product, whose absorbance at 335 to 345nm can be measured with a plate reader or spectrophotometer.
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›