Thermo Scientific Zeba 40K and 7K Desalt Resin Performance
Sample volumes of 100µL of different compounds were processed with 0.5mL of two Zeba Resins, as well as with two 30K-50K MWCO resins from Suppliers G and B. The Zeba Resins removed 85-100% of DTT, NaCl and biotin.
Sample volumes of 100µL of different compounds were processed with 0.5mL of two Zeba Resins, as well as with two 30K-50K MWCO resins from Suppliers G and B. The Zeba Resins removed 85-100% of DTT, NaCl and biotin.
Related Products
Related Images