Software Downloads
High Resolution Melting (HRM) Software V3.0.1
High Resolution Melting Software v3.0.1 is an all-new and improved tool for High Resolution Melting (HRM) analysis. HRM analysis is a post-PCR analysis method used to identify variation in nucleic acid sequences. The method is based on detecting small differences in PCR melting (dissociation) curves. It is enabled by high-brightness, dsDNA-binding dyes used in conjunction with real-time PCR instrumentation that has precise temperature ramp control, advanced data capture capabilities, and access to software designed specifically for HRM analysis. High Resolution Melting Software v3.0.1 brings HRM to the StepOne™ and StepOnePlus™ Real-Time PCR Systems, and the 7500 Fast Real-Time PCR System, and contains features that make fast and accurate HRM results more accessible than ever.
To download a FREE 30-day trial of High Resolution Melting Software v3.0.1, click here.
For users of the 7500 Fast Real-Time PCR System who have already purchased High Resolution Melting Software v3.0, click here to download the HRM v3.0.1 patch.
PC Download High Resolution Melting Software v3.0.1 (1 license)
PC Download High Resolution Melting Software v3.0.1 (10 licenses)