Search
The desktop instrument operating software enables users to operate QuantStudio 12K Flex instrument. The software can be run with security, audit, and e-signature (SAE) enabled.
The companion OpenArray Accufill software is also available to assist with OpenArray plate preparation.
Design & Analysis 2 (DA2) software enables users to set up run templates, open completed run files to analyze data, and review results in an integrated and streamlined fashion. The software provides a suite of secondary analysis modules for specific qPCR applications. The software also supports OpenArray image visualization for quality control and troubleshooting.
While DA2 software can be run with security, audit, and e-signature (SAE) enabled, this feature does not apply to the QuantStudio 12K system.
The latest Design and Analysis software allows users to enjoy all the same features as Design & Analysis 2 (DA2) software while offering additional features and enhancements. Users can now access the software in any one of three deployment options: classic cesktop experience (available in Windows or MAC), cloud flexibility, and on-premises server-client configuration for multi-user access via the security, audit, and e-signature (SAE) enabled environment*. Utilize multiplate analysis capabilities with both genotyping and copy number variation analysis modules. Diomni Design and Analysis (RUO) Software v3.0 is the feature-rich solution that connects all the steps of your qPCR workflows into a single software experience.
*SAE is not available with the desktop version of software. SAE is only available with the on-premises version of software.
For Research Use Only. Not for use in diagnostic procedures.