Search
We are dedicated to your success. Get back on track. View our expert recommendations for commonly encountered problem scenarios with your DNA methylation analysis experiments.
View the relevant questions below:
Beginning your experiment?
Visit our
When there is very little or no methylated DNA, MBD protein also binds non-methylated DNA to some extent. So it is important to follow the right protocol when using low DNA input. The product manual has different protocols specified for different DNA input amounts.
Ensure that the DNA used for bisulfite conversion is pure. If particulate matter is present after adding the CT Conversion Reagent, then centrifuge the material at high speed and perform the conversion with the clear supernatant. Also, ensure that all of the liquid is at the bottom and not in the cap or walls of the PCR tube before performing the conversion reaction.
For next-generation sequencing troubleshooting help, please visit the troubleshooting pages within our Next-Generation Sequencing Support Center.
Please go here for troubleshooting help.
Here are some recommendations:
For the 7500 Fast Real-Time PCR System, if the software version is below v2.0.4, the compatible HRM software is v2.0.1. If the 7500 Fast Real-Time PCR System has been upgraded to software version 2.0.4 or above, then you will need to get HRM Software version 3.0.1.
There are a few possibilities:
For Research Use Only. Not for use in diagnostic procedures.