Search
In analytical labs where details matter most—when productivity is measured in microns and minutes—turn to the Thermo Scientific Nicolet RaptIR or RaptIR+ FTIR Microscope for detailed answers.
These research-grade microscopes help you rapidly locate and identify trace materials, inclusions, impurities, microparticles, and determine the distribution of materials within a sample with incredible clarity and precision. Its clean, adaptive user interface provides increased flexibility and usability for all users, from specialists to novices. The Nicolet RaptIR and RaptIR+ FTIR Microscopes provide answers quickly, regardless of industry or application.
Expanding your options, the RaptIR+ adds the versatility of user-swappable detectors offering extended-range operation or liquid-nitrogen-free operation. When adapted to interface with a Nicolet iS50 FTIR Spectrometer, the RaptIR+ enables a wide range of new analysis possibilities, including TGA-IR, GC-IR, NIR, FT-Raman, and much more. For a more cost-conscious choice, the RaptIR+ can also adapt to the Nicolet Apex FTIR Spectrometer.
From the mosaic feature to infrared capability and a seamless user experience, the Nicolet RaptIR and RaptIR+ FTIR Microscopes are full of technological innovations that help specialists and non-specialists alike.
Nicolet RaptIR+ FTIR Microscope Walkthrough
The Nicolet RaptIR and RaptIR+ FTIR Microscopes are always coupled to a Thermo Scientific Nicolet FTIR Spectrometer. Using it with the Thermo Scientific Nicolet iS50 FTIR Spectrometer provides a broader array of capabilities—including FT-Raman, TGA-IR, NIR, far-infrared, and diamond ATR—for addressing complex analytical challenges. This combination will help you find answers more quickly, whether you are studying powders, liquids, gases, or micron-sized particulates.
| Nicolet RaptIR+ FTIR Microscope | Nicolet RaptIR FTIR Microscope | |
|---|---|---|
Visible resolution | 1 micron | 1 micron |
Infrared resolution | 5 micron | 5 micron |
Multiple detectors | Swappable, MCT-A, MCT-B, InGaAs | Fixed, MCT-A, MCT-B |
Spectrometer option | Nicolet iS50 FTIR Spectrometer (right or left side) Nicolet Apex FTIR Spectrometer (right side) | Nicolet iS50 FTIR Spectrometer (right side) |
Objectives | 4X or 40X visible, 15X or grazing angle infrared | 4X or 40X visible, 15X or grazing angle infrared |
Polarizers | Motorized, visible and IR | Motorized, visible and IR |
The Nicolet RaptIR FTIR Microscope's adaptable design is suitable for scientists across all industries and skill levels. It has been engineered with the latest technology to ensure you get sharp images and rapid, accurate analysis at every step of your workflow.
The Nicolet RaptIR+ FTIR Microscope automatically ensures you can use it anywhere, anytime.
Load samples up to 40 millimeters thick and 5 kilograms in mass with the open access and heavy-duty stage.
The high-resolution camera and eyepieces display clear images, and the adaptive UI and comfortable joystick provide a nearly effortless user experience.
The 4x and 15x objectives enable you to get a clear view of your entire sample and focus in on the smallest detail.
Cover a large area faster than ever with the microscope's infrared capability and examine your sample with up to 10 spectra per second at a resolution greater than 5 microns.
Paradigm Software 64-bit operating system and UI simplify your entire analytical process.
The microscope facilitates the transmission of data into a wide variety of formal and fully customizable formats to send to your stakeholders.
The Nicolet RaptIR+ FTIR Microscope automatically ensures you can use it anywhere, anytime.
Load samples up to 40 millimeters thick and 5 kilograms in mass with the open access and heavy-duty stage.
The high-resolution camera and eyepieces display clear images, and the adaptive UI and comfortable joystick provide a nearly effortless user experience.
The 4x and 15x objectives enable you to get a clear view of your entire sample and focus in on the smallest detail.
Cover a large area faster than ever with the microscope's infrared capability and examine your sample with up to 10 spectra per second at a resolution greater than 5 microns.
Paradigm Software 64-bit operating system and UI simplify your entire analytical process.
The microscope facilitates the transmission of data into a wide variety of formal and fully customizable formats to send to your stakeholders.
The versatility of the RaptIR+/RaptIR allows you to analyze across a wide range of applications:
See it fast. Identify it faster. Watch this video to experience the Nicolet RaptIR FTIR Microscope—designed with enhanced precision and agility to help streamline analysis and generate actionable results with amazing speed.
Thermo Scientific OMNIC Paradigm Software allows you to swiftly navigate and analyze complex samples. It employs a database format for collecting data, utilizes the full power of the Windows 10 operating system, and collects impressively large maps to enable you to collect multiple regions of interest.
The innovative hardware features of the Nicolet RaptIR+ FTIR Microscope are complemented by the advantages of the OMNIC Paradigm Desktop Software. It provides users with clear guidance on how to extract information needed from the target.
The software boasts a clean, adaptive user interface (UI), database security, floating tool bar, 64-bit system, and easily generated workflows to automate data collection and analysis sequences.
10 micron beads embedded in an epoxy
Fibers finely resolved and analyzed
Incredible detail of ink on a plastic surface
Examine entire tablets
Gain insight about API distribution
Colors of complex images
Forensic sample details - auto part contamination
Forensic sample details - paint chip analysis
Analyze the environmental impact of microplastics
The software’s clean, adaptive user interface allows specialists and novice users alike to obtain the data they’re looking for. Workflow automation, combined with the ability to select multiple regions of interest in a sample for collection in one pass, helps save time and reduce errors. Autofocus and automated illumination allow for quick and easy data collection. Fully customizable microparticles analysis workflow allows users of varying experience to click, get images, and move forward, choosing what they want to show in tailored reports.
See for yourself in these short videos how easy it is to use the Nicolet RaptIR+/RaptIR FTIR Microscope to find a sample, collect a mosaic, or collect a background and map:
Find your sample fast using software
Collect the background and an image map of your sample
Collect a high-resolution mosaic using software
Analysis of a high-res multi-layer film
Contact us today to learn how you too can obtain answers in minutes and microns with the Nicolet RaptIR+/RaptIR FTIR Microscopes and Thermo Scientific OMNIC Paradigm Software.
Please complete the form below and we will contact you shortly.