Search
| Component | Amount per HeLa Cell | Amount per E. coli cell |
|---|---|---|
| Total dry weight | 400 pg | 0.4 pg |
| Total DNA | 15 pg | 0.017 pg |
| Total RNA | 30 pg | 0.10 pg |
| Total protein | 300 pg | 0.2 pg |
| Cytoplasmic ribosomes | 4 x 106 | 3 x 104 |
| Cytoplasmic tRNA molecules | 6 x 107 | 4 x 105 |
| Cytoplasmic mRNA molecules | 7 x 105 | 4 x 103 |