Search
Maintain compliance with official dioxin and PCB methods, whether you are in the U.S., EU, or Japan. Achieve confidence and robustness in your quantitative routine analysis of dioxins and persistent organic pollutants (POPs) with a Thermo Scientific DFS Magnetic Sector GC-HRMS and Thermo Scientific Orbitrap Exploris GC S Mass Spectrometer.
For over a decade, the global scientific community has chosen the DFS Magnetic Sector GC-HRMS for its superior performance in routine quantitative analysis of dioxins and POPs. Today, the DFS Magnetic Sector GC-HRMS continues to set the gold standard in providing robust performance and high-confidence results.
Based on magnetic sector technology, the DFS Magnetic Sector GC-HRMS performs dioxin and POPs routine analysis, delivering ultimate confidence no matter the sample type or matrix. It features a large-volume ion source and high-capacity turbo pump for maximum sensitivity and robustness.
Both food import-export control and environmental analyses require compliance with numerous government and analytical methods. The DFS Magnetic Sector GC-HRMS is the only GC-MS system compliant with existing official methods and regulations, including U.S. EPA, EU, and JIS methods.
High-linearity detection and matrix-independent calibrations combined with automated mass calibration for each analysis run provide confidence in your analytical results. Additionally, the system displays the best dioxin installation specifications during each system installation (based on 2378-TCDD with 4 sigma noise definition) and makes LOD/LOQ calculations using S/N real-time detection limits calculated for each sample analysis.
The DFS Magnetic Sector GC-HRMS features lock mass trace for constant monitoring of instrument stability and sample matrix effect in compliance with official methods, such as EPA 1613. Additionally, mass resolution is documented in a log file for each analysis, providing proof for stable selectivity.
The flexible configuration of the DFS Magnetic Sector GC-HRMS offers a unique dual GC configuration with the option to install up to 4 columns. The system can operate with different reference gases, e.g., PFK, FC43, etc.
The ion source can be exchanged without breaking vacuum for quick maintenance. Additionally, a separation valve enables venting of the ion source housing when necessary.
Experience a sensitive, versatile, future-ready platform for regulated and evolving POPs workflows, powered by Orbitrap technology, for confidence in every result to support high-throughput testing.
Delivering confidence and robustness, the DFS Magnetic Sector GC-HRMS meets your current and future needs for worldwide compliance in dioxins and POPs analysis.
Curious to know 6 facts about Magnetic Sector Mass Spectrometers and Thermo Scientific DFS Magnetic Sector GC-HRMS for Dioxin analysis? Watch the series and let us know more facts!
Meet with the team at NofaLab Laboratories (Rotterdam), where the team explains their dioxin analysis workflow with Magnetic Sector GC-HRMS and how important is fast turnaround time for their customers.
Follow the DFS Magnetic Sector GC-HRMS Product Team in their visit at Pacific Rim Lab, a very productive Dioxins and POPs lab.
Curious to know 6 facts about Magnetic Sector Mass Spectrometers and Thermo Scientific DFS Magnetic Sector GC-HRMS for Dioxin analysis? Watch the series and let us know more facts!
Meet with the team at NofaLab Laboratories (Rotterdam), where the team explains their dioxin analysis workflow with Magnetic Sector GC-HRMS and how important is fast turnaround time for their customers.
Follow the DFS Magnetic Sector GC-HRMS Product Team in their visit at Pacific Rim Lab, a very productive Dioxins and POPs lab.
The Orbitrap Exploris GC 240 mass spectrometer provides comprehensive characterization of samples in a single analysis.
The Thermo Scientific TRACE 1610 gas chromatograph provides versatility and exceptional GC and GC-MS performance.
Our comprehensive portfolio of GC columns ensures that we have the right column to meet your analytical needs.
* Required field