Product References
T cells drive negative feedback mechanisms in cancer associated fibroblasts, promoting expression of co-inhibitory ligands, CD73 and IL-27 in non-small cell lung cancer.
Oncoimmunology
O'Connor RA,Chauhan V,Mathieson L,Titmarsh H,Koppensteiner L,Young I,Tagliavini G,Dorward DA,Prost S,Dhaliwal K,Wallace WA,Akram AR
12-5888-42 was used in Flow Cytometry to study co-inhibitory ligands in T cell immunosuppression in non-small cell lung cancer.
Mon Aug 02 00:00:00 EDT 2021
Inhibition of PI3Kδ Differentially Regulates Poly I:C- and Human Metapneumovirus-Induced PD-L1 and PD-L2 Expression in Human Bronchial Epithelial Cells.
Frontiers in immunology
Ogawa T,Kan-O K,Shiota A,Fujita A,Ishii Y,Fukuyama S,Matsumoto K
12-5888-42 was used in Flow Cytometry to suggest that inhibition of PI3Kδ differentially regulates PD-L1 and PD-L2 expression in bronchial epithelial cells during viral infections.
Mon Feb 14 00:00:00 EST 2022
Inhibition of PI3Kδ Differentially Regulates Poly I:C- and Human Metapneumovirus-Induced PD-L1 and PD-L2 Expression in Human Bronchial Epithelial Cells.
Frontiers in immunology
Ogawa T,Kan-O K,Shiota A,Fujita A,Ishii Y,Fukuyama S,Matsumoto K
12-5888-42 was used in Flow Cytometry to suggest that inhibition of PI3Kδ differentially regulates PD-L1 and PD-L2 expression in bronchial epithelial cells during viral infections.
Mon Feb 14 00:00:00 EST 2022
Osteosarcoma cell intrinsic PD-L2 signals promote invasion and metastasis via the RhoA-ROCK-LIMK2 and autophagy pathways.
Cell death & disease
Ren T,Zheng B,Huang Y,Wang S,Bao X,Liu K,Guo W
Published figure using CD273 (B7-DC) monoclonal antibody (Product # 12-5888-42) in Flow Cytometry
Mon Mar 18 00:00:00 EDT 2019
TLR9 Mediated Tumor-Stroma Interactions in Human Papilloma Virus (HPV)-Positive Head and Neck Squamous Cell Carcinoma Up-Regulate PD-L1 and PD-L2.
Frontiers in immunology
Baruah P,Bullenkamp J,Wilson POG,Lee M,Kaski JC,Dumitriu IE
Published figure using CD273 (B7-DC) monoclonal antibody (Product # 12-5888-42) in Flow Cytometry
Wed Oct 14 00:00:00 EDT 2020
EWS-FLI1 low Ewing sarcoma cells demonstrate decreased susceptibility to T-cell-mediated tumor cell apoptosis.
Oncotarget
Bailey KM,Julian CM,Klinghoffer AN,Bernard H,Lucas PC,McAllister-Lucas LM
12-5888-42 was used in Flow Cytometry to discover that EWS-FLI1 low Ewing sarcoma cells are more resistant to T-cell mediated apoptosis due to upregulation of PD-L1 and PD-L2.
Tue May 21 00:00:00 EDT 2019
PD-1 axis expression in musculoskeletal tumors and antitumor effect of nivolumab in osteosarcoma model of humanized mouse.
Journal of hematology & oncology
Zheng B,Ren T,Huang Y,Sun K,Wang S,Bao X,Liu K,Guo W
Published figure using CD273 (B7-DC) monoclonal antibody (Product # 12-5888-42) in Flow Cytometry
Tue Feb 06 00:00:00 EST 2018
PD-L1 expression in medulloblastoma: an evaluation by subgroup.
Oncotarget
Martin AM,Nirschl CJ,Polanczyk MJ,Bell WR,Nirschl TR,Harris-Bookman S,Phallen J,Hicks J,Martinez D,Ogurtsova A,Xu H,Sullivan LM,Meeker AK,Raabe EH,Cohen KJ,Eberhart CG,Burger PC,Santi M,Taube JM,Pardoll DM,Drake CG,Lim M
12-5888 was used in Flow cytometry/Cell sorting to evaluate the expression of PD-L1 in human medulloblastoma subgroups.
Tue Apr 10 00:00:00 EDT 2018
Butyrate Conditions Human Dendritic Cells to Prime Type 1 Regulatory T Cells via both Histone Deacetylase Inhibition and G Protein-Coupled Receptor 109A Signaling.
Frontiers in immunology
Kaisar MMM,Pelgrom LR,van der Ham AJ,Yazdanbakhsh M,Everts B
12-5888 was used in Flow cytometry/Cell sorting to study the effect of short chain fatty acids on human dendritic cells and how they affect their capacity to prime and polarise T cell responses.
Wed Nov 20 00:00:00 EST 2019
CD64-directed microtubule associated protein tau kills leukemic blasts ex vivo.
Oncotarget
Mladenov R,Hristodorov D,Cremer C,Gresch G,Grieger E,Schenke L,Klose D,Amoury M,Woitok M,Jost E,Brümmendorf TH,Fendel R,Fischer R,Stein C,Thepen T,Barth S
12-5888 was used in Flow cytometry/Cell sorting to investigate how leukaemia may be treated by CD64-directed MAP tau.
Tue Oct 11 00:00:00 EDT 2016
Paucity of PD-L1 expression in prostate cancer: innate and adaptive immune resistance.
Prostate cancer and prostatic diseases
Martin AM,Nirschl TR,Nirschl CJ,Francica BJ,Kochel CM,van Bokhoven A,Meeker AK,Lucia MS,Anders RA,DeMarzo AM,Drake CG
12-5888 was used in Flow cytometry/Cell sorting to investigate whether prostate cancer cells were capable of expressing PD-L1.
Tue Dec 01 00:00:00 EST 2015
Cervical (pre)neoplastic microenvironment promotes the emergence of tolerogenic dendritic cells via RANKL secretion.
Oncoimmunology
Demoulin SA,Somja J,Duray A,Guénin S,Roncarati P,Delvenne PO,Herfs MF,Hubert PM
12-5888 was used in Flow cytometry/Cell sorting to investigate how the emergence of tolerogenic dendritic cells via RANKL secretion is promoted by cervical (pre)neoplastic microenvironment.
Mon Jun 01 00:00:00 EDT 2015
HIV-1 Tat protein induces PD-L1 (B7-H1) expression on dendritic cells through tumor necrosis factor alpha- and toll-like receptor 4-mediated mechanisms.
Journal of virology
Planès R,BenMohamed L,Leghmari K,Delobel P,Izopet J,Bahraoui E
12-5888 was used in Flow cytometry/Cell sorting to investigate the mechanisms by which HIV-1 induces upregulation of coinhibitory molecules, showing that the HIV-1 Tat protein induces PD-L1 expression on dendritic cells.
Sun Jun 01 00:00:00 EDT 2014
Brugia malayi microfilariae induce a regulatory monocyte/macrophage phenotype that suppresses innate and adaptive immune responses.
PLoS neglected tropical diseases
O'Regan NL,Steinfelder S,Venugopal G,Rao GB,Lucius R,Srikantam A,Hartmann S
12-5888 was used in Flow cytometry/Cell sorting to show that monocytes stimulated with Brugia malayi microfilarial lysate develop a defined regulatory phenotype.
Wed Oct 01 00:00:00 EDT 2014
Programmed death ligand 1 is expressed by non-hodgkin lymphomas and inhibits the activity of tumor-associated T cells.
Clinical cancer research : an official journal of the American Association for Cancer Research
Andorsky DJ,Yamada RE,Said J,Pinkus GS,Betting DJ,Timmerman JM
12-5888 was used in Flow cytometry/Cell sorting to characterise the spectrum of PD-L1 expression in non-Hodgkin lymphomas and evaluate the role of PD-L1 in suppressing tumour-associated T-cell responses.
Fri Jul 01 00:00:00 EDT 2011
Phagocytosis, a potential mechanism for myeloid-derived suppressor cell regulation of CD8+ T cell function mediated through programmed cell death-1 and programmed cell death-1 ligand interaction.
Journal of immunology (Baltimore, Md. : 1950)
Kim YJ,Park SJ,Broxmeyer HE
12-5888 was used in Flow cytometry/Cell sorting to identify a potential mechanism by which IFN-γ-producing CD8(+) T cells are tolerised after type 1 immune responses to chronic virus or tumour.
Thu Sep 01 00:00:00 EDT 2011
Transfer of regulatory properties from tolerogenic to proinflammatory dendritic cells via induced autoreactive regulatory T cells.
Journal of immunology (Baltimore, Md. : 1950)
Kleijwegt FS,Laban S,Duinkerken G,Joosten AM,Koeleman BP,Nikolic T,Roep BO
12-5888 was used in Flow cytometry/Cell sorting to study the mechanism by which infectious tolerance applies to human dendritic cells.
Thu Dec 15 00:00:00 EST 2011
Increase of programmed death-1-expressing intratumoral CD8 T cells predicts a poor prognosis for nasopharyngeal carcinoma.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc
Hsu MC,Hsiao JR,Chang KC,Wu YH,Su IJ,Jin YT,Chang Y
12-5888 was used in Immunofluorescence to propose that PD-1 expression in CD8 cells reflects the selective suppression of cytotoxic lymphocytes in the tumor microenvironment.
Fri Oct 01 00:00:00 EDT 2010
Major depletion of plasmacytoid dendritic cells in HIV-2 infection, an attenuated form of HIV disease.
PLoS pathogens
Cavaleiro R,Baptista AP,Soares RS,Tendeiro R,Foxall RB,Gomes P,Victorino RM,Sousa AE
12-5888 was used in Flow cytometry/Cell sorting to understand the contribution of plasmacytoid dendritic cell biology to HIV/AIDS pathogenesis.
Sun Nov 01 00:00:00 EDT 2009
PD-1 is a regulator of NY-ESO-1-specific CD8+ T cell expansion in melanoma patients.
Journal of immunology (Baltimore, Md. : 1950)
Fourcade J,Kudela P,Sun Z,Shen H,Land SR,Lenzner D,Guillaume P,Luescher IF,Sander C,Ferrone S,Kirkwood JM,Zarour HM
Published figure using CD273 (B7-DC) monoclonal antibody (Product # 12-5888-42) in Flow Cytometry
Fri May 01 00:00:00 EDT 2009
TLR-mediated induction of negative regulatory ligands on dendritic cells.
Journal of molecular medicine (Berlin, Germany)
Gröschel S,Piggott KD,Vaglio A,Ma-Krupa W,Singh K,Goronzy JJ,Weyand CM
12-5888 was used in Flow cytometry/Cell sorting to demonstrate that dendritic cell stimulation through the TLR3 ligand dsRNA [poly(I:C)] limits CD4 T-cell proliferation.
Tue Apr 01 00:00:00 EDT 2008
No evidence for dualism in function and receptors: PD-L2/B7-DC is an inhibitory regulator of human T cell activation.
European journal of immunology
Pfistershammer K,Klauser C,Pickl WF,Stöckl J,Leitner J,Zlabinger G,Majdic O,Steinberger P
Mon May 01 00:00:00 EDT 2006