Product References
A Matrix-Assisted Laser Desorption/Ionization Time-of-Flight Assay Identifies Nilotinib as an Inhibitor of Inflammation in Acute Myeloid Leukemia.
Journal of medicinal chemistry
Marín-Rubio JL,Peltier-Heap RE,Dueñas ME,Heunis T,Dannoura A,Inns J,Scott J,Simpson AJ,Blair HJ,Heidenreich O,Allan JM,Watt JE,Martin MP,Saxty B,Trost M
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 14-0541-82) in Flow Cytometry
Thu Sep 22 00:00:00 EDT 2022
ICAM-1 and ICAM-2 Are Differentially Expressed and Up-Regulated on Inflamed Pulmonary Epithelium, but Neither ICAM-2 nor LFA-1: ICAM-1 Are Required for Neutrophil Migration Into the Airways In Vivo.
Frontiers in immunology
Chong DLW,Rebeyrol C,José RJ,Williams AE,Brown JS,Scotton CJ,Porter JC
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 14-0541-82) in Immunohistochemistry
Mon Oct 25 00:00:00 EDT 2021
ICAM-1 and ICAM-2 Are Differentially Expressed and Up-Regulated on Inflamed Pulmonary Epithelium, but Neither ICAM-2 nor LFA-1: ICAM-1 Are Required for Neutrophil Migration Into the Airways In Vivo.
Frontiers in immunology
Chong DLW,Rebeyrol C,José RJ,Williams AE,Brown JS,Scotton CJ,Porter JC
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 14-0541-82) in Immunohistochemistry
Mon Oct 25 00:00:00 EDT 2021
Neutrophil-endothelial interactions of murine cells is not a good predictor of their interactions in human cells.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Soroush F,Tang Y,Mustafa O,Sun S,Yang Q,Kilpatrick LE,Kiani MF
14-0541-81 was used in Immunocytochemistry to determine whether the response of murine cells to inflammatory activation or anti-inflammatory treatment is predictive of the response in human cells.
Sat Feb 01 00:00:00 EST 2020
L-Selectin/CD62L is a Key Driver of Non-Alcoholic Steatohepatitis in Mice and Men.
Cells
Drescher HK,Schippers A,Rosenhain S,Gremse F,Bongiovanni L,Bruin A,Eswaran S,Gallage SU,Pfister D,Szydlowska M,Heikenwalder M,Weiskirchen S,Wagner N,Trautwein C,Weiskirchen R,Kroy DC
14-0541 was used in Immunohistochemistry-immunofluorescence to investigate the functional role of CD62L in NASH in humans as well as in two mouse models of steatohepatitis.
Wed Apr 29 00:00:00 EDT 2020
Conserved regulation of RNA processing in somatic cell reprogramming.
BMC genomics
Kanitz A,Syed AP,Kaji K,Zavolan M
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 14-0541-82) in Flow Cytometry
Thu Jan 31 00:00:00 EST 2019
PAI1 mediates fibroblast-mast cell interactions in skin fibrosis.
The Journal of clinical investigation
Pincha N,Hajam EY,Badarinath K,Batta SPR,Masudi T,Dey R,Andreasen P,Kawakami T,Samuel R,George R,Danda D,Jacob PM,Jamora C
14-0541 was used in Immunohistochemistry to identify roles for plasminogen activator inhibitor type 1 in the multifactorial program of fibrogenesis that expand its functional repertoire beyond its canonical role in plasmin-dependent processes.
Tue May 01 00:00:00 EDT 2018
PAI1 mediates fibroblast-mast cell interactions in skin fibrosis.
The Journal of clinical investigation
Pincha N,Hajam EY,Badarinath K,Batta SPR,Masudi T,Dey R,Andreasen P,Kawakami T,Samuel R,George R,Danda D,Jacob PM,Jamora C
14-0541 was used in Immunohistochemistry to identify roles for plasminogen activator inhibitor type 1 in the multifactorial program of fibrogenesis that expand its functional repertoire beyond its canonical role in plasmin-dependent processes.
Tue May 01 00:00:00 EDT 2018
Low-Dose Radiotherapy Has No Harmful Effects on Key Cells of Healthy Non-Inflamed Joints.
International journal of molecular sciences
Deloch L,Rückert M,Fietkau R,Frey B,Gaipl US
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 14-0541-82) in Flow Cytometry
Tue Oct 16 00:00:00 EDT 2018
Constitutively Active SMAD2/3 Are Broad-Scope Potentiators of Transcription-Factor-Mediated Cellular Reprogramming.
Cell stem cell
Ruetz T,Pfisterer U,Di Stefano B,Ashmore J,Beniazza M,Tian TV,Kaemena DF,Tosti L,Tan W,Manning JR,Chantzoura E,Ottosson DR,Collombet S,Johnsson A,Cohen E,Yusa K,Linnarsson S,Graf T,Parmar M,Kaji K
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 14-0541-82) in Flow Cytometry
Thu Dec 07 00:00:00 EST 2017
Endogenous TNFα orchestrates the trafficking of neutrophils into and within lymphatic vessels during acute inflammation.
Scientific reports
Arokiasamy S,Zakian C,Dilliway J,Wang W,Nourshargh S,Voisin MB
14-0541 was used in Immunofluorescence to demonstrate a new role for TNFα as a key regulator of neutrophil trafficking into and within the lymphatic system.
Mon Mar 13 00:00:00 EDT 2017
A robust in vitro model for trans-lymphatic endothelial migration.
Scientific reports
Xiong Y,Brinkman CC,Famulski KS,Mongodin EF,Lord CJ,Hippen KL,Blazar BR,Bromberg JS
14-0541 was used in Flow cytometry/Cell sorting to establish an in vitro system to evaluate lymphatic Trans-endothelial migration for various cell types across primary mouse and human lymphatic endothelial cells.
Tue May 09 00:00:00 EDT 2017
Palmitoyl acyltransferase DHHC21 mediates endothelial dysfunction in systemic inflammatory response syndrome.
Nature communications
Beard RS,Yang X,Meegan JE,Overstreet JW,Yang CG,Elliott JA,Reynolds JJ,Cha BJ,Pivetti CD,Mitchell DA,Wu MH,Deschenes RJ,Yuan SY
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 14-0541-82) in Immunofluorescence
Thu Sep 22 00:00:00 EDT 2016
Inhibition of the TNF Family Cytokine RANKL Prevents Autoimmune Inflammation in the Central Nervous System.
Immunity
Guerrini MM,Okamoto K,Komatsu N,Sawa S,Danks L,Penninger JM,Nakashima T,Takayanagi H
14-0541-82 was used in Flow Cytometry to show that receptor activator of nuclear factor-κB (NF-κB) ligand (RANKL) on T cells regulates C-C type chemokine ligand 20 (CCL20) production by astrocytes and T cell localization in the CNS.
Tue Dec 15 00:00:00 EST 2015
Reprogramming Roadblocks Are System Dependent.
Stem cell reports
Chantzoura E,Skylaki S,Menendez S,Kim SI,Johnsson A,Linnarsson S,Woltjen K,Chambers I,Kaji K
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 14-0541-82) in Flow Cytometry
Tue Sep 08 00:00:00 EDT 2015
Neutrophil crawling in capillaries; a novel immune response to Staphylococcus aureus.
PLoS pathogens
Harding MG,Zhang K,Conly J,Kubes P
14-0541 was used in Blocking experiments to elucidate the importance of neutrophils in protection against MRSA-induced soft tissue infections.
Wed Oct 01 00:00:00 EDT 2014
Dietary gluten increases natural killer cell cytotoxicity and cytokine secretion.
European journal of immunology
Larsen J,Dall M,Antvorskov JC,Weile C,Engkilde K,Josefsen K,Buschard K
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 14-0541-82) in Flow Cytometry
Wed Oct 01 00:00:00 EDT 2014
High-resolution analysis with novel cell-surface markers identifies routes to iPS cells.
Nature
O'Malley J,Skylaki S,Iwabuchi KA,Chantzoura E,Ruetz T,Johnsson A,Tomlinson SR,Linnarsson S,Kaji K
Published figure using CD54 (ICAM-1) monoclonal antibody (Product # 14-0541-82) in Flow Cytometry
Thu Jul 04 00:00:00 EDT 2013
ICAM-1-mediated leukocyte adhesion is critical for the activation of endothelial LSP1.
American journal of physiology. Cell physiology
Hossain M,Qadri SM,Su Y,Liu L
14-0541 was used in Functional assays to study the role of endothelial leukocyte-specific protein 1 in leukocyte adhesion through ICAM-1.
Wed May 01 00:00:00 EDT 2013
Pericytes support neutrophil subendothelial cell crawling and breaching of venular walls in vivo.
The Journal of experimental medicine
Proebstl D,Voisin MB,Woodfin A,Whiteford J,D'Acquisto F,Jones GE,Rowe D,Nourshargh S
14-0541 was used in Immunofluorescence to identify previously unknown roles for pericytes in neutrophil transmigration in vivo and leukocyte trafficking into sites of inflammation.
Mon Jun 04 00:00:00 EDT 2012
Cutting edge: Humanized nano-proresolving medicines mimic inflammation-resolution and enhance wound healing.
Journal of immunology (Baltimore, Md. : 1950)
Norling LV,Spite M,Yang R,Flower RJ,Perretti M,Serhan CN
14-0541 was used in Flow cytometry/Cell sorting to study the temporal generation and properties of endogenous microparticles produced in evolving self-limited inflammatory exudates in vivo.
Sun May 15 00:00:00 EDT 2011
Requirement of interaction between mast cells and skin dendritic cells to establish contact hypersensitivity.
PloS one
Otsuka A,Kubo M,Honda T,Egawa G,Nakajima S,Tanizaki H,Kim B,Matsuoka S,Watanabe T,Nakae S,Miyachi Y,Kabashima K
14-0541 was used in Neutralization experiments to investigate the role of mast cells in contact hypersensitivity, showing that interaction between mast cells and skin dendritic cells is required.
Thu Jan 26 00:00:00 EST 2012
Steroid-resistant lymphatic remodeling in chronically inflamed mouse airways.
The American journal of pathology
Yao LC,Baluk P,Feng J,McDonald DM
14-0541 was used in Immunohistochemistry to suggest that lymphatic remodelling after the initial inflammatory response may influence later inflammatory episodes.
Mon Mar 01 00:00:00 EST 2010
Annexin-1-deficient dendritic cells acquire a mature phenotype during differentiation.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Huggins A,Paschalidis N,Flower RJ,Perretti M,D'Acquisto F
14-0541 was used in Flow cytometry/Cell sorting to suggest that the inhibition of Anx-A1 expression in dendritic cells may be useful to modulate the adaptive immune response in pathogen-induced immune diseases.
Wed Apr 01 00:00:00 EDT 2009
Microvascular endothelial cells exhibit optimal aspect ratio for minimizing flow resistance.
Annals of biomedical engineering
Sumagin R,Brown CW,Sarelius IH,King MR
14-0541 was used in Immunofluorescence to reveal that the spacing of endethelial nuclei in venules is similar to a predicted optimal configuration to minimise flow resistance.
Tue Apr 01 00:00:00 EDT 2008
Murine cerebral malaria development is independent of toll-like receptor signaling.
The American journal of pathology
Togbe D,Schofield L,Grau GE,Schnyder B,Boissay V,Charron S,Rose S,Beutler B,Quesniaux VF,Ryffel B
14-0541 was used in Immunohistochemistry to investigate innate immune activation by malarial molecular motifs and the potential underlying signalling mechanisms.
Tue May 01 00:00:00 EDT 2007
Prevention of lipopolysaccharide-induced microangiopathy by gp49B1: evidence for an important role for gp49B1 expression on neutrophils.
The Journal of experimental medicine
Zhou JS,Friend DS,Feldweg AM,Daheshia M,Li L,Austen KF,Katz HR
14-0541 was used in Blocking experiments to investigate the role of gp49B1 in neutrophils, showing that gp49B1 expression on neutrophils suppresses LPS-induced microangiopathy.
Mon Oct 20 00:00:00 EDT 2003