Product References
Re-Visiting Antioxidant Therapy in Murine Advanced Atherosclerosis with Brussels Chicory, a Typical Vegetable in Mediterranean Diets.
Nutrients
Li Q,Du Y,Xiang P,Chen G,Qian X,Li S,Mao Y,Ling W,Wang D
14-0681-82 was used in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) to investigate whether Brussels chicory can stabilize advanced plaques in the aorta via improving oxidative stress.
Mon Feb 06 00:00:00 EST 2023
Re-Visiting Antioxidant Therapy in Murine Advanced Atherosclerosis with Brussels Chicory, a Typical Vegetable in Mediterranean Diets.
Nutrients
Li Q,Du Y,Xiang P,Chen G,Qian X,Li S,Mao Y,Ling W,Wang D
14-0681-82 was used in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) to investigate whether Brussels chicory can stabilize advanced plaques in the aorta via improving oxidative stress.
Mon Feb 06 00:00:00 EST 2023
Glycolytic System in Axons Supplement Decreased ATP Levels after Axotomy of the Peripheral Nerve.
eNeuro
Takenaka T,Ohnishi Y,Yamamoto M,Setoyama D,Kishima H
14-0681-82 was used in Immunohistochemistry-immunofluorescence to suggest that glycolytic system, both in Schwann cells and axons, is the main source of maintaining ATP levels in the distal nerve stump.
Wed Mar 01 00:00:00 EST 2023
P2Y6 receptor-dependent microglial phagocytosis of synapses mediates synaptic and memory loss in aging.
Aging cell
Dundee JM,Puigdellívol M,Butler R,Cockram TOJ,Brown GC
14-0681-82 was used in Immunohistochemistry to show that inactivation of P2Y6 R decreases microglial phagocytosis of isolated synapses (synaptosomes) and synaptic loss in neuronal-glial co-cultures.
Wed Feb 01 00:00:00 EST 2023
Single-cell transcriptomic dissection of the cellular and molecular events underlying the triclosan-induced liver fibrosis in mice.
Military Medical Research
Bai YM,Yang F,Luo P,Xie LL,Chen JH,Guan YD,Zhou HC,Xu TF,Hao HW,Chen B,Zhao JH,Liang CL,Dai LY,Geng QS,Wang JG
14-0681-82 was used in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) to provide the first comprehensive single-cell atlas of mouse livers in response to TCS and delineate the key cellular and molecular processes involved in TCS-induced hepatotoxicity and fibrosis.
Wed Feb 22 00:00:00 EST 2023
Adventitial macrophage accumulation impairs perivascular nerve function in mesenteric arteries with inflammatory bowel disease.
Frontiers in physiology
Grunz EA,Jones BW,Lateef OM,Sen S,Wilkinson K,Joshi T,Boerman EM
14-0681-82 was used in Immunohistochemistry-immunofluorescence to find perivascular nerve function is impaired in mesenteric arteries with Inflammatory bowel disease.
Sat Jul 01 00:00:00 EDT 2023
Melanoma-Secreted Amyloid Beta Suppresses Neuroinflammation and Promotes Brain Metastasis.
Cancer discovery
Kleffman K,Levinson G,Rose IVL,Blumenberg LM,Shadaloey SAA,Dhabaria A,Wong E,Galán-Echevarría F,Karz A,Argibay D,Von Itter R,Floristán A,Baptiste G,Eskow NM,Tranos JA,Chen J,Vega Y Saenz de Miera EC,Call M,Rogers R,Jour G,Wadghiri YZ,Osman I,Li YM,Mathews P,DeMattos RB,Ueberheide B,Ruggles KV,Liddelow SA,Schneider RJ,Hernando E
14-0681-82 was used in Immunohistochemistry to reveal a novel mechanistic connection between brain metastasis and Alzheimer's disease, two previously unrelated pathologies; establish Aβ as a promising therapeutic target for brain metastasis; and demonstrate suppression of neuroinflammation as a critical feature of metastatic adaptation to the brain parenchyma.
Mon May 02 00:00:00 EDT 2022
An enriched environment re-establishes metabolic homeostasis by reducing obesity-induced inflammation.
Disease models & mechanisms
Díaz de León-Guerrero S,Salazar-León J,Meza-Sosa KF,Valle-Garcia D,Aguilar-León D,Pedraza-Alva G,Pérez-Martínez L
14-0681-82 was used in Immunohistochemistry to explore whether an enriched environment is capable of restoring energy balance in obese mice that previously presented metabolic alterations.
Wed Jun 01 00:00:00 EDT 2022
Myeloid cell-specific ablation of Runx2 gene exacerbates post-infarct cardiac remodeling.
Scientific reports
Tomimatsu M,Matsumoto K,Ashizuka M,Kumagai S,Tanaka S,Nakae T,Yokota K,Kominami S,Kajiura R,Okuzaki D,Motooka D,Shiraishi A,Abe T,Matsuda H,Okada Y,Maeda M,Seno S,Obana M,Fujio Y
Published figure using CD68 monoclonal antibody (Product # 14-0681-82) in Immunohistochemistry
Wed Oct 05 00:00:00 EDT 2022
Exposure to Microbial Metabolite Butyrate Prolongs the Survival Time and Changes the Growth Pattern of Human Papillomavirus 16 E6/E7-Immortalized Keratinocytes in Vivo.
The American journal of pathology
Li M,McGhee EM,Shinno L,Lee K,Lin YL
14-0681-82 was used in Immunohistochemistry to show that butyrate treatment changed the property of HPV16 E6/E7-immortalized keratinocytes.
Fri Oct 01 00:00:00 EDT 2021
Sulfiredoxin-1 attenuates injury and inflammation in acute pancreatitis through the ROS/ER stress/Cathepsin B axis.
Cell death & disease
He J,Ma M,Li D,Wang K,Wang Q,Li Q,He H,Zhou Y,Li Q,Hou X,Yang L
14-0681-82 was used in Immunohistochemistry and Immunocytochemistry to show that Srxn1 is a protective target for AP by attenuating acinar injury and inflammation through the ROS/ER stress/Cathepsin B axis.
Thu Jun 17 00:00:00 EDT 2021
Sulfiredoxin-1 attenuates injury and inflammation in acute pancreatitis through the ROS/ER stress/Cathepsin B axis.
Cell death & disease
He J,Ma M,Li D,Wang K,Wang Q,Li Q,He H,Zhou Y,Li Q,Hou X,Yang L
14-0681-82 was used in Immunohistochemistry and Immunocytochemistry to show that Srxn1 is a protective target for AP by attenuating acinar injury and inflammation through the ROS/ER stress/Cathepsin B axis.
Thu Jun 17 00:00:00 EDT 2021
Sulfiredoxin-1 attenuates injury and inflammation in acute pancreatitis through the ROS/ER stress/Cathepsin B axis.
Cell death & disease
He J,Ma M,Li D,Wang K,Wang Q,Li Q,He H,Zhou Y,Li Q,Hou X,Yang L
14-0681-82 was used in Immunohistochemistry and Immunocytochemistry to show that Srxn1 is a protective target for AP by attenuating acinar injury and inflammation through the ROS/ER stress/Cathepsin B axis.
Thu Jun 17 00:00:00 EDT 2021
Environmental Enrichment Induces Meningeal Niche Remodeling through TrkB-Mediated Signaling.
International journal of molecular sciences
Zorzin S,Corsi A,Ciarpella F,Bottani E,Dolci S,Malpeli G,Pino A,Amenta A,Fumagalli GF,Chiamulera C,Bifari F,Decimo I
14-0681-82 was used in Immunohistochemistry to analyse the effects of environmental enrichment exposure on neural precursors distribution in mouse brain meninges.
Fri Oct 01 00:00:00 EDT 2021
Environmental Enrichment Induces Meningeal Niche Remodeling through TrkB-Mediated Signaling.
International journal of molecular sciences
Zorzin S,Corsi A,Ciarpella F,Bottani E,Dolci S,Malpeli G,Pino A,Amenta A,Fumagalli GF,Chiamulera C,Bifari F,Decimo I
14-0681-82 was used in Immunohistochemistry to analyse the effects of environmental enrichment exposure on neural precursors distribution in mouse brain meninges.
Fri Oct 01 00:00:00 EDT 2021
Endothelial Heparan Sulfate Mediates Hepatic Neutrophil Trafficking and Injury during Staphylococcus aureus Sepsis.
mBio
Golden GJ,Toledo AG,Marki A,Sorrentino JT,Morris C,Riley RJ,Spliid C,Chen Q,Cornax I,Lewis NE,Varki N,Le D,Malmström J,Karlsson C,Ley K,Nizet V,Esko JD
Published figure using CD68 monoclonal antibody (Product # 14-0681-82) in Immunocytochemistry
Tue Oct 26 00:00:00 EDT 2021
TREM2 activation on microglia promotes myelin debris clearance and remyelination in a model of multiple sclerosis.
Acta neuropathologica
Cignarella F,Filipello F,Bollman B,Cantoni C,Locca A,Mikesell R,Manis M,Ibrahim A,Deng L,Benitez BA,Cruchaga C,Licastro D,Mihindukulasuriya K,Harari O,Buckland M,Holtzman DM,Rosenthal A,Schwabe T,Tassi I,Piccio L
14-0681 was used in Immunohistochemistry-immunofluorescence to demonstrate that TREM2 was highly expressed on myelin-laden phagocytes in active demyelinating lesions in the CNS of subjects with multiple sclerosis.
Thu Oct 01 00:00:00 EDT 2020
Paeoniflorin relieves LPS-induced inflammatory pain in mice by inhibiting NLRP3 inflammasome activation via transient receptor potential vanilloid 1.
Journal of leukocyte biology
Yin N,Gao Q,Tao W,Chen J,Bi J,Ding F,Wang Z
14-0681-82 was used in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) to suggest that PF can be as a potential pharmacological agent for inflammatory pain and thus deserves more attention and further investigation.
Wed Jul 01 00:00:00 EDT 2020
Rhinovirus impairs the immune response of alveolar macrophages to facilitate Streptococcus pneumonia infection.
Pathogens and disease
Wei H,Zhao H,Li R,Yang F,Wu Y
14-0681 was used in Immunohistochemistry to investigate how human rhinovirus (HRV) would accelerate Streptococcus pneumonia infection.
Wed Apr 01 00:00:00 EDT 2020
Identification of ILK as a critical regulator of VEGFR3 signalling and lymphatic vascular growth.
The EMBO journal
Urner S,Planas-Paz L,Hilger LS,Henning C,Branopolski A,Kelly-Goss M,Stanczuk L,Pitter B,Montanez E,Peirce SM,Mäkinen T,Lammert E
14-0681-82 was used in Immunohistochemistry to investigate how vascular endothelial growth factor receptor-3 (VEGFR3) signalling is attenuated to prevent lymphatic vascular overgrowth and ensure proper lymph vessel development.
Tue Jan 15 00:00:00 EST 2019
Sam68 impedes the recovery of arterial injury by augmenting inflammatory response.
Journal of molecular and cellular cardiology
Han S,Xu S,Zhou J,Qiao A,Boriboun C,Ma W,Li H,Biyashev D,Yang L,Zhang E,Liu Q,Jiang S,Zhao TC,Krishnamurthy P,Zhang C,Richard S,Qiu H,Zhang J,Qin G
14-0681 was used in Immunohistochemistry-immunofluorescence to dissect the molecular mechanism by which Sam68 regulates NF-κB signaling and its functional significance in vascular injury.
Sun Dec 01 00:00:00 EST 2019
Sympathetic Neuronal Activation Triggers Myeloid Progenitor Proliferation and Differentiation.
Immunity
Vasamsetti SB,Florentin J,Coppin E,Stiekema LCA,Zheng KH,Nisar MU,Sembrat J,Levinthal DJ,Rojas M,Stroes ESG,Kim K,Dutta P
14-0681 was used in Immunohistochemistry to demonstrate that catecholamines produced by leukocytes and sympathetic nerve termini promote GMP proliferation and myeloid cell development.
Tue Jul 17 00:00:00 EDT 2018
Macrophage-Derived Neuropilin-2 Exhibits Novel Tumor-Promoting Functions.
Cancer research
Roy S,Bag AK,Dutta S,Polavaram NS,Islam R,Schellenburg S,Banwait J,Guda C,Ran S,Hollingsworth MA,Singh RK,Talmadge JE,Muders MH,Batra SK,Datta K
14-0681-82 was used in Immunohistochemistry to demonstrate that NRP2 is expressed during macrophage differentiation, is induced by tumour cells, and regulates phagocytosis in macrophages.
Mon Oct 01 00:00:00 EDT 2018
Macrophage-Derived Neuropilin-2 Exhibits Novel Tumor-Promoting Functions.
Cancer research
Roy S,Bag AK,Dutta S,Polavaram NS,Islam R,Schellenburg S,Banwait J,Guda C,Ran S,Hollingsworth MA,Singh RK,Talmadge JE,Muders MH,Batra SK,Datta K
14-0681-82 was used in Immunohistochemistry to demonstrate that NRP2 is expressed during macrophage differentiation, is induced by tumour cells, and regulates phagocytosis in macrophages.
Mon Oct 01 00:00:00 EDT 2018
Heparanase regulates the M1 polarization of renal macrophages and their crosstalk with renal epithelial tubular cells after ischemia/reperfusion injury.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Masola V,Zaza G,Bellin G,Dall'Olmo L,Granata S,Vischini G,Secchi MF,Lupo A,Gambaro G,Onisto M
14-0681 was used in Western Blotting to show that heparanase regulates macrophage polarisation as well as renal damage and repair after ischemia/reperfusion injury.
Thu Feb 01 00:00:00 EST 2018
Heparanase regulates the M1 polarization of renal macrophages and their crosstalk with renal epithelial tubular cells after ischemia/reperfusion injury.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Masola V,Zaza G,Bellin G,Dall'Olmo L,Granata S,Vischini G,Secchi MF,Lupo A,Gambaro G,Onisto M
14-0681 was used in Western Blotting to show that heparanase regulates macrophage polarisation as well as renal damage and repair after ischemia/reperfusion injury.
Thu Feb 01 00:00:00 EST 2018
Effects of ganoderic acid A on lipopolysaccharide-induced proinflammatory cytokine release from primary mouse microglia cultures.
Experimental and therapeutic medicine
Chi B,Wang S,Bi S,Qin W,Wu D,Luo Z,Gui S,Wang D,Yin X,Wang F
Published figure using CD68 monoclonal antibody (Product # 14-0681-82) in Flow Cytometry
Mon Jan 01 00:00:00 EST 2018
Suppression of LRRC19 promotes cutaneous wound healing in pressure ulcers in mice.
Organogenesis
Sun J,Wang Z,Wang X
14-0681-82 was used in Immunohistochemistry to support that LRRC19 could be a potential therapeutic target for pressure ulcers.
Tue Jan 02 00:00:00 EST 2018
iPSC-Derived Macrophages Effectively Treat Pulmonary Alveolar Proteinosis in Csf2rb-Deficient Mice.
Stem cell reports
Mucci A,Lopez-Rodriguez E,Hetzel M,Liu S,Suzuki T,Happle C,Ackermann M,Kempf H,Hillje R,Kunkiel J,Janosz E,Brennig S,Glage S,Bankstahl JP,Dettmer S,Rodt T,Gohring G,Trapnell B,Hansen G,Trapnell C,Knudsen L,Lachmann N,Moritz T
Published figure using CD68 monoclonal antibody (Product # 14-0681-82) in Immunohistochemistry
Tue Sep 11 00:00:00 EDT 2018
Streamlining volumetric multi-channel image cytometry using hue-saturation-brightness-based surface creation.
Communications biology
Tan Y,Li JLY,Goh CC,Lee BTK,Kwok IWH,Ng WJ,Evrard M,Poidinger M,Tey HL,Ng LG
14-0681 was used in Flow cytometry/Cell sorting to streamline volumetric multi-channel image cytometry using hue-saturation-brightness-based surface creation.
Sat Jan 11 00:00:00 EST 2020
Streamlining volumetric multi-channel image cytometry using hue-saturation-brightness-based surface creation.
Communications biology
Tan Y,Li JLY,Goh CC,Lee BTK,Kwok IWH,Ng WJ,Evrard M,Poidinger M,Tey HL,Ng LG
14-0681 was used in Flow cytometry/Cell sorting to streamline volumetric multi-channel image cytometry using hue-saturation-brightness-based surface creation.
Sat Jan 11 00:00:00 EST 2020
Streamlining volumetric multi-channel image cytometry using hue-saturation-brightness-based surface creation.
Communications biology
Tan Y,Li JLY,Goh CC,Lee BTK,Kwok IWH,Ng WJ,Evrard M,Poidinger M,Tey HL,Ng LG
14-0681 was used in Flow cytometry/Cell sorting to streamline volumetric multi-channel image cytometry using hue-saturation-brightness-based surface creation.
Sat Jan 11 00:00:00 EST 2020
L2hgdh Deficiency Accumulates l-2-Hydroxyglutarate with Progressive Leukoencephalopathy and Neurodegeneration.
Molecular and cellular biology
Ma S,Sun R,Jiang B,Gao J,Deng W,Liu P,He R,Cui J,Ji M,Yi W,Yang P,Wu X,Xiong Y,Qiu Z,Ye D,Guan KL
Published figure using CD68 monoclonal antibody (Product # 14-0681-82) in Immunofluorescence
Sat Apr 15 00:00:00 EDT 2017
Epicardium is required for cardiac seeding by yolk sac macrophages, precursors of resident macrophages of the adult heart.
Developmental biology
Stevens SM,von Gise A,VanDusen N,Zhou B,Pu WT
Published figure using CD68 monoclonal antibody (Product # 14-0681-82) in Immunofluorescence
Sun May 15 00:00:00 EDT 2016
CD68 acts as a major gateway for malaria sporozoite liver infection.
The Journal of experimental medicine
Cha SJ,Park K,Srinivasan P,Schindler CW,van Rooijen N,Stins M,Jacobs-Lorena M
14-0681-82 was used in Immunocytochemistry to examine the molecular determinants of malarial sporozoite-Kupffer cell interactions.
Mon Aug 24 00:00:00 EDT 2015
Breaking immune tolerance by targeting Foxp3(+) regulatory T cells mitigates Alzheimer's disease pathology.
Nature communications
Baruch K,Rosenzweig N,Kertser A,Deczkowska A,Sharif AM,Spinrad A,Tsitsou-Kampeli A,Sarel A,Cahalon L,Schwartz M
Published figure using CD68 monoclonal antibody (Product # 14-0681-82) in Immunofluorescence
Tue Aug 18 00:00:00 EDT 2015
Sialoadhesin deficiency does not influence the severity of lupus nephritis in New Zealand black x New Zealand white F1 mice.
Arthritis research & therapy
Kidder D,Richards HE,Lyons PA,Crocker PR
14-0681 was used in Immunohistochemistry on frozen tissues to investigate a potential causative role of Sialoadhesin in lupus.
Fri Nov 01 00:00:00 EDT 2013
CD11b⁺, Ly6G⁺ cells produce type I interferon and exhibit tissue protective properties following peripheral virus infection.
PLoS pathogens
Fischer MA,Davies ML,Reider IE,Heipertz EL,Epler MR,Sei JJ,Ingersoll MA,Rooijen NV,Randolph GJ,Norbury CC
14-0681 was used in Flow cytometry/Cell sorting to demonstrate that innate immune effector cells contain a peripheral poxvirus infection, preventing systemic spread of the virus.
Tue Nov 01 00:00:00 EDT 2011