Product References
Correlation between E-cadherin expression and tumor grade, proliferation, microvascular density, and apoptosis in canine cutaneous squamous cell carcinoma.
Canadian journal of veterinary research = Revue canadienne de recherche veterinaire
Gupta A,Al-Dissi A
14-5699-82 was used in Immunohistochemistry to examine the expression of E-cadherin in canine cutaneous squamous cell carcinoma (SCC) and determined its association with tumor grade (TG), proliferation index (PI), apoptosis index (AI), and intra-tumoral microvascular density (iMVD) in archived samples.
Sun Jan 01 00:00:00 EST 2023
GABP couples oncogene signaling to telomere regulation in TERT promoter mutant cancer.
Cell reports
McKinney AM,Mathur R,Stevers NO,Molinaro AM,Chang SM,Phillips JJ,Costello JF
14-5699-82 was used in Flow Cytometry to study whether the role of epidermal growth factor receptor (EGFR) amplification to drive proliferation and telomerase reverse transcriptase promoter (TERTp) mutations underlying telomerase reactivation through recruitment of GA-binding protein (GABP) are functionally connected in cancer.
Tue Sep 20 00:00:00 EDT 2022
Acetyl-CoA Synthetase 2: A Critical Linkage in Obesity-Induced Tumorigenesis in Myeloma.
Cell metabolism
Li Z,Liu H,He J,Wang Z,Yin Z,You G,Wang Z,Davis RE,Lin P,Bergsagel PL,Manasanch EE,Wong STC,Esnaola NF,Chang JC,Orlowski RZ,Yi Q,Yang J
14-5699-82 was used in Immunohistochemistry to demonstrate a key impact for obesity-induced acetyl-CoA synthetase 2 on the progression of myeloma.
Tue Jan 05 00:00:00 EST 2021
Protein/AS01B vaccination elicits stronger, more Th2-skewed antigen-specific human T follicular helper cell responses than heterologous viral vectors.
Cell reports. Medicine
Nielsen CM,Ogbe A,Pedroza-Pacheco I,Doeleman SE,Chen Y,Silk SE,Barrett JR,Elias SC,Miura K,Diouf A,Bardelli M,Dabbs RA,Barfod L,Long CA,Haynes BF,Payne RO,Minassian AM,Bradley T,Draper SJ,Borrow P
Published figure using Ki-67 monoclonal antibody (Product # 14-5699-82) in Flow Cytometry
Tue Mar 16 00:00:00 EDT 2021
Microbial exposure during early human development primes fetal immune cells.
Cell
Mishra A,Lai GC,Yao LJ,Aung TT,Shental N,Rotter-Maskowitz A,Shepherdson E,Singh GSN,Pai R,Shanti A,Wong RMM,Lee A,Khyriem C,Dutertre CA,Chakarov S,Srinivasan KG,Shadan NB,Zhang XM,Khalilnezhad S,Cottier F,Tan ASM,Low G,Chen P,Fan Y,Hor PX,Lee AKM,Choolani M,Vermijlen D,Sharma A,Fuks G,Straussman R,Pavelka N,Malleret B,McGovern N,Albani S,Chan JKY,Ginhoux F
14-5699-82 was used in Immunocytochemistry, Immunohistochemistry (Paraffin) to indicate selective presence of live microbes in fetal organs during the 2nd trimester of gestation and have broader implications toward the establishment of immune competency and priming before birth.
Thu Jun 24 00:00:00 EDT 2021
Effects of Tyrosine Kinase Inhibitor-masitinib Mesylate on Canine Mammary Tumour Cell Lines.
Journal of veterinary research
Ustun-Alkan F,Bakırel T,Üstüner O,Anlas C,Cinar S,Yıldırım F,Gürel A
14-5699-82 was used in Immunocytochemistry, Immunohistochemistry (Paraffin) to investigate the antiproliferative effect of masitinib in CMT cells and its possible mechanisms of action.
Wed Sep 01 00:00:00 EDT 2021
T cells drive negative feedback mechanisms in cancer associated fibroblasts, promoting expression of co-inhibitory ligands, CD73 and IL-27 in non-small cell lung cancer.
Oncoimmunology
O'Connor RA,Chauhan V,Mathieson L,Titmarsh H,Koppensteiner L,Young I,Tagliavini G,Dorward DA,Prost S,Dhaliwal K,Wallace WA,Akram AR
14-5699-82 was used in Flow Cytometry to study co-inhibitory ligands in T cell immunosuppression in non-small cell lung cancer.
Mon Aug 02 00:00:00 EDT 2021
Circulating Tumor Cells Exhibit Metastatic Tropism and Reveal Brain Metastasis Drivers.
Cancer discovery
Klotz R,Thomas A,Teng T,Han SM,Iriondo O,Li L,Restrepo-Vassalli S,Wang A,Izadian N,MacKay M,Moon BS,Liu KJ,Ganesan SK,Lee G,Kang DS,Walmsley CS,Pinto C,Press MF,Lu W,Lu J,Juric D,Bardia A,Hicks J,Salhia B,Attenello F,Smith AD,Yu M
14-5699-82 was used in Immunohistochemistry (Paraffin) to demonstrate that ex vivo-cultured CTCs from 4 patients with breast cancer showed organotropism, revealing molecular features that allow a subset of CTCs to enter and grow in the brain.
Wed Jan 01 00:00:00 EST 2020
Single residue in CD28-costimulated CAR-T cells limits long-term persistence and antitumor durability.
The Journal of clinical investigation
Guedan S,Madar A,Casado-Medrano V,Shaw C,Wing A,Liu F,Young RM,June CH,Posey AD
Published figure using Ki-67 monoclonal antibody (Product # 14-5699-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2020
Human Liver Memory CD8+ T Cells Use Autophagy for Tissue Residence.
Cell reports
Swadling L,Pallett LJ,Diniz MO,Baker JM,Amin OE,Stegmann KA,Burton AR,Schmidt NM,Jeffery-Smith A,Zakeri N,Suveizdyte K,Froghi F,Fusai G,Rosenberg WM,Davidson BR,Schurich A,Simon AK,Maini MK
Published figure using Ki-67 monoclonal antibody (Product # 14-5699-82) in Flow Cytometry
Tue Jan 21 00:00:00 EST 2020
Functional Study of the Retrotransposon-Derived Human PEG10 Protease.
International journal of molecular sciences
Golda M,Mótyán JA,Mahdi M,Tőzsér J
Published figure using Ki-67 monoclonal antibody (Product # 14-5699-82) in Flow Cytometry
Tue Mar 31 00:00:00 EDT 2020
Niraparib activates interferon signaling and potentiates anti-PD-1 antibody efficacy in tumor models.
Scientific reports
Wang Z,Sun K,Xiao Y,Feng B,Mikule K,Ma X,Feng N,Vellano CP,Federico L,Marszalek JR,Mills GB,Hanke J,Ramaswamy S,Wang J
Published figure using Ki-67 monoclonal antibody (Product # 14-5699-82) in Flow Cytometry
Tue Feb 12 00:00:00 EST 2019
Tissue-resident memory T cells are epigenetically cytotoxic with signs of exhaustion in human urinary bladder cancer.
Clinical and experimental immunology
Hartana CA,Ahlén Bergman E,Broomé A,Berglund S,Johansson M,Alamdari F,Jakubczyk T,Huge Y,Aljabery F,Palmqvist K,Holmström B,Glise H,Riklund K,Sherif A,Winqvist O
14-5699 was used in Flow cytometry/Cell sorting to identify TRM cells as potential new targets for cancer immunotherapy.
Mon Oct 01 00:00:00 EDT 2018
Involvement of TWEAK and the NF-κB signaling pathway in lupus nephritis.
Experimental and therapeutic medicine
Sun F,Teng J,Yu P,Li W,Chang J,Xu H
Published figure using Ki-67 monoclonal antibody (Product # 14-5699-82) in Flow Cytometry
Thu Mar 01 00:00:00 EST 2018
Highly multiplexed immunofluorescence imaging of human tissues and tumors using t-CyCIF and conventional optical microscopes.
eLife
Lin JR,Izar B,Wang S,Yapp C,Mei S,Shah PM,Santagata S,Sorger PK
Published figure using Ki-67 monoclonal antibody (Product # 14-5699-82) in Flow Cytometry
Wed Jul 11 00:00:00 EDT 2018
Small-Vessel Vasculopathy Due to Aberrant Autophagy in LAMP-2 Deficiency.
Scientific reports
Nguyen HT,Noguchi S,Sugie K,Matsuo Y,Nguyen CTH,Koito H,Shiojima I,Nishino I,Tsukaguchi H
Published figure using Ki-67 monoclonal antibody (Product # 14-5699-82) in Flow Cytometry
Tue Feb 20 00:00:00 EST 2018
Streptomyces Levis ABRIINW111 Inhibits SW480 Cells Growth by Apoptosis Induction.
Advanced pharmaceutical bulletin
Faramarzian Azimi Maragheh B,Fatourachi P,Mohammadi SM,Valipour B,Behtari M,Dehnad A,Nozad Charoudeh H
Published figure using Ki-67 monoclonal antibody (Product # 14-5699-82) in Flow Cytometry
Thu Nov 01 00:00:00 EDT 2018
Core Transcription Factors, MicroRNAs, and Small Molecules Drive Transdifferentiation of Human Fibroblasts Towards The Cardiac Cell Lineage.
Scientific reports
Christoforou N,Chakraborty S,Kirkton RD,Adler AF,Addis RC,Leong KW
14569982 was used in immunocytochemistry to describe the transdifferentiation of human dermal fibroblasts towards the cardiac cell lineage
Tue Jan 10 00:00:00 EST 2017
Core Transcription Factors, MicroRNAs, and Small Molecules Drive Transdifferentiation of Human Fibroblasts Towards The Cardiac Cell Lineage.
Scientific reports
Christoforou N,Chakraborty S,Kirkton RD,Adler AF,Addis RC,Leong KW
14569982 was used in immunocytochemistry to describe the transdifferentiation of human dermal fibroblasts towards the cardiac cell lineage
Tue Jan 10 00:00:00 EST 2017
S100A6 Regulates Endothelial Cell Cycle Progression by Attenuating Antiproliferative Signal Transducers and Activators of Transcription 1 Signaling.
Arteriosclerosis, thrombosis, and vascular biology
Lerchenmüller C,Heißenberg J,Damilano F,Bezzeridis VJ,Krämer I,Bochaton-Piallat ML,Hirschberg K,Busch M,Katus HA,Peppel K,Rosenzweig A,Busch H,Boerries M,Most P
Published figure using Ki-67 monoclonal antibody (Product # 14-5699-82) in Immunocytochemistry
Thu Sep 01 00:00:00 EDT 2016
S100A6 Regulates Endothelial Cell Cycle Progression by Attenuating Antiproliferative Signal Transducers and Activators of Transcription 1 Signaling.
Arteriosclerosis, thrombosis, and vascular biology
Lerchenmüller C,Heißenberg J,Damilano F,Bezzeridis VJ,Krämer I,Bochaton-Piallat ML,Hirschberg K,Busch M,Katus HA,Peppel K,Rosenzweig A,Busch H,Boerries M,Most P
Published figure using Ki-67 monoclonal antibody (Product # 14-5699-82) in Immunocytochemistry
Thu Sep 01 00:00:00 EDT 2016
Interleukin-7 and Toll-like receptor 7 induce synergistic B cell and T cell activation.
PloS one
Bikker A,Kruize AA,van der Wurff-Jacobs KM,Peters RP,Kleinjan M,Redegeld F,de Jager W,Lafeber FP,van Roon JA
Published figure using Ki-67 monoclonal antibody (Product # 14-5699-82) in Flow Cytometry
Sat Feb 07 00:00:00 EST 2015
Somatostatin expression in human hair follicles and its potential role in immune privilege.
The Journal of investigative dermatology
Breitkopf T,Lo BK,Leung G,Wang E,Yu M,Carr N,Zloty D,Cowan B,Shapiro J,McElwee KJ
14-5699 was used in Flow cytometry/Cell sorting to demonstrate and quantify the functional immune privilege properties apparent in human hair follicles.
Mon Jul 01 00:00:00 EDT 2013
Biliary tree stem cells, precursors to pancreatic committed progenitors: evidence for possible life-long pancreatic organogenesis.
Stem cells (Dayton, Ohio)
Wang Y,Lanzoni G,Carpino G,Cui CB,Dominguez-Bendala J,Wauthier E,Cardinale V,Oikawa T,Pileggi A,Gerber D,Furth ME,Alvaro D,Gaudio E,Inverardi L,Reid LM
Published figure using Ki-67 monoclonal antibody (Product # 14-5699-82) in Immunofluorescence
Sun Sep 01 00:00:00 EDT 2013
Dll4-Notch signaling in Flt3-independent dendritic cell development and autoimmunity in mice.
The Journal of experimental medicine
Billiard F,Lobry C,Darrasse-Jèze G,Waite J,Liu X,Mouquet H,DaNave A,Tait M,Idoyaga J,Leboeuf M,Kyratsous CA,Burton J,Kalter J,Klinakis A,Zhang W,Thurston G,Merad M,Steinman RM,Murphy AJ,Yancopoulos GD,Aifantis I,Skokos D
14-5699 was used in Flow cytometry/Cell sorting to investigate how Dll4-Notch signaling affects pro-T cell fate and thymic DC development, showing that it is a novel Flt3-independent pathway for DC development and autoimmunity.
Mon May 07 00:00:00 EDT 2012